Skip to Content
Merck

Skip To

P7136

Pyrazinecarboxamide

Synonym(s):

Pyrazinamide, Pyrazinoic acid amide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C5H5N3O
CAS Number:
Molecular Weight:
123.11
UNSPSC Code:
41116107
NACRES:
NA.85
PubChem Substance ID:
EC Number:
202-717-6
Beilstein/REAXYS Number:
112306
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

mp

189-191 °C (lit.)

antibiotic activity spectrum

mycobacteria

mode of action

cell membrane | interferes

SMILES string

NC(=O)c1cnccn1

InChI

1S/C5H5N3O/c6-5(9)4-3-7-1-2-8-4/h1-3H,(H2,6,9)

InChI key

IPEHBUMCGVEMRF-UHFFFAOYSA-N

Application

Pyrazinamide is used therapeutically as an antitubercular agent. Pyrazinamide is used to form polymeric copper complexes, create pyrazine carboxamide scaffolds useful as FXs inhibitors, and as a component of mycobacteria identification kits. It is used to study liver toxicity prevention [1] and mechanisms of resistance [2].

Biochem/physiol Actions

The active moiety of pyrazinamide is pyrazinoic acid (POA). POA is thought to disrupt membrane energetics and inhibit membrane transport function at acid pH in Mycobacterium tuberculosis. Iron enhances the antituberculous activity of pyrazinamide [3]. Pyrazinamide and its analogs have been shown to inhibit the activity of purified FAS I.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A76958P56100PHR1576
mode of action

cell membrane | interferes

mode of action

-

mode of action

-

mode of action

-

antibiotic activity spectrum

mycobacteria

antibiotic activity spectrum

-

antibiotic activity spectrum

-

antibiotic activity spectrum

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

300

form

powder

form

crystals

form

powder

form

-

mp

189-191 °C (lit.)

mp

118-120 °C (lit.)

mp

222-225 °C (dec.) (lit.)

mp

189-191 °C (lit.)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
P7136-25G04061834394389
P7136-10G04061834394372
P7136-100G04061834394365

Questions

Reviews

No rating value

Active Filters