Skip to Content
Merck

Skip To

P6887

Pepsin from porcine gastric mucosa

lyophilized powder, ≥3,200 units/mg protein

Synonym(s):

Pepsin A, Pepsin from hog stomach

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
eCl@ss:
42010127
EC Number:
232-629-3
NACRES:
NA.54
MDL number:
EC Number:
Specific activity:
≥3,200 units/mg protein
Biological source:
Porcine gastric mucosa
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Porcine gastric mucosa

Quality Level

grade

Proteomics Grade

form

lyophilized powder

specific activity

≥3,200 units/mg protein

mol wt

35 kDa

impurities

salt, essentially free

color

white to off-white

solubility

deionized water: soluble 10 mg/mL, 10 mM HCl: soluble 4.0 mg/mL (Cold)

UniProt accession no.

application(s)

diagnostic assay manufacturing

shipped in

wet ice

storage temp.

−20°C

Gene Information

Application

Pepsin from Sigma has been used along with other enzymes for the determination of enzyme-resistant starch (RS) in bread. It has also been used to simulate in vitro gastrointestinal digestion of pea or whey protein isolates.[1]
Pepsin cleavage can be used to produce F(ab′)2 fragments of antibodies. pepsin at www.sigma-aldrich.com/enzymeexplorer.
Pepsin is a peptidase used to digest proteins and is commonly used in the preparation of Fab fragments from antibodies. Pepsin, from porcine gastric mucosa, has been used to hydrolyze dry cervical samples in mice. Product P6887 is provided as a lyophilized powder and has been used to digest protein during dietary fiber analysis[2].

Biochem/physiol Actions

Unlike many other peptidases, pepsin hydrolyzes only peptide bonds, not amide or ester linkages. The cleavage specificity includes peptides with an aromatic acid on either side of the peptide bond, especially if the other residue is also an aromatic or a dicarboxylic amino acid. Increased susceptibility to hydrolysis occurs if there is a sulfur-containing amino acid close to the peptide bond, which has an aromatic amino acid. Pepsin will also preferentially cleave at the carboxyl side of phenylalanine and leucine, and to a lesser extent at the carboxyl side of glutamic acid residues. It does not cleave at valine, alanine, or glycine linkages. Z-L-tyrosyl-L-phenylalanine, Z-L-glutamyl-L-tyrosine, or Z-L-methionyl-L-tyrosine may be used as substrates for pepsin digestion. Pepsin is inhibited by several phenylalanine-containing peptides.[3]
Preferential cleavage: hydrophobic and aromatic residues in P1 and P1′ postitions. Cleaves Phe-Val, Gln-His, Glu-Ala, Ala-Leu, Leu-Tyr, Tyr-Leu, Gly-Phe, Phe-Phe and Phe-Tyr bonds in the β chain of insulin

Analysis Note

Optimum pH is 2-4. Active in 4 M urea and 3 M guanidine HCl. Stable at 60 °C. Pepsin is irreversibly inactivated at pH 8.0 - 8.5.
Protein determined by E1%/280

Other Notes

One unit will produce a ΔA280 of 0.001 per min at pH 2.0 at 37°C, measured as TCA-soluble products using hemoglobin as substrate. (Final volume = 16 ml. Light path = 1 cm.)
View more information on pepsin at www.sigma-aldrich.com/enzymeexplorer.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P7012SRE0001P7000
biological source

Porcine gastric mucosa

biological source

-

biological source

-

biological source

Porcine gastric mucosa

grade

Proteomics Grade

grade

-

grade

-

grade

-

specific activity

≥3,200 units/mg protein

specific activity

≥2,500 units/mg protein (E1%/280)

specific activity

≥3200 units/mg protein

specific activity

≥250 units/mg solid

Gene Information

pig ... LOC396892(396892)

Gene Information

pig ... LOC396892(396892)

Gene Information

pig ... LOC396892(396892)

Gene Information

pig ... LOC396892(396892)

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

powder


Still not finding the right product?

Explore all of our products under Pepsin from porcine gastric mucosa


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Enzyme-resistant starch. III. The quality of straight-dough bread containing varying levels of enzyme-resistant starch.
Eerlingen, R. C., et al.
Cereal Chem., 71(2), 165-169 (1994)
J A Marlett et al.
The American journal of clinical nutrition, 50(3), 479-485 (1989-09-01)
The effects of different methods on the distribution of total neutral sugars (TNS), uronic acids (UA), and beta-glucans (beta G) between the soluble (S) and insoluble (I) fractions of dietary fiber (DF) were determined for peas, kidney beans, oat bran
Enzymes of Molecular Biology
Sweeney, P.J., and Walker, J.M.
Methods in Molecular Biology, 290-291 (1993)



Global Trade Item Number

SKUGTIN
P6887-5G04061835558551
P6887-10G04061835558520
P6887-1G04061835558537
P6887-250MG04061835558544

Questions

Reviews

No rating value

Active Filters