Skip to Content
Merck

Skip To

P6874

Anti-p53 antibody, Mouse monoclonal

clone DO-1, purified from hybridoma cell culture

Synonym(s):

Anti-BCC7, Anti-BMFS5, Anti-LFS1, Anti-P53, Anti-TRP53

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
DO-1, monoclonal
Application:
ARR, ELISA (i), FACS, ICC, IHC, IP, WB
Citations:
27
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DO-1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~53 kDa

species reactivity

human

concentration

~2 mg/mL

technique(s)

flow cytometry: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, indirect ELISA: suitable, microarray: suitable, western blot: 0.2-0.4 μg/mL using total cell extract of A431 cells

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TP53(7157)

General description

Monoclonal Anti-p53 (mouse IgG2a isotype) is derived from hybridoma DO-1 produced by the fusion of mouse myeloma cells (SP2 cells) and splenocytes from BALB/c mice immunized with recombinant human wild type p53. The p53 gene is located on human chromosome 17p. The gene codes for a tumor suppressor protein that is expressed in normal tissues.

Immunogen

recombinant human wild type p53. The epitope resides between amino acids 20-25 of human p53.

Application

Anti-p53 antibody, Mouse monoclonal has been used in:
  • semi-dry western blotting
  • immunoblotting[1]
  • immunofluorescence[1]
  • enzyme-linked immunosorbent assay (ELISA)
  • immunocytochemistry
  • immunohistochemistry
  • immunoprecipitation
  • flow cytometry
  • chromatin immunoprecipitation (ChIP) assay

Biochem/physiol Actions

Mouse anti-(p53) antibody reacts specifically with (p53) if human.
The p53 tumor suppressor protein is important in the cellular response to DNA damage and other genomic aberrations. Elevation of p53 protein induces the transcriptional activation of multiple genes, including p21awf1. Wild-type p53 is shown to be a sequence-specific transcription factor, directly interacting with various cellular and viral proteins. Intact p53 function is essential for the maintenance of the non-tumorogenic phenotype of cells.p53 has a pivotal role in suppressing the development of cancer, cell cycle regulation and modulation of CAK kinase activity.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P6749P8999P5813
Gene Information

human ... TP53(7157)

Gene Information

human ... TP53(7157)

Gene Information

human ... TP53(7157)

Gene Information

human ... TP53(7157)

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

ascites fluid

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

clone

DO-1, monoclonal

clone

PAb1801, monoclonal

clone

DO-7, monoclonal

clone

BP53-12, monoclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



D V Bulavin et al.
The EMBO journal, 18(23), 6845-6854 (1999-12-03)
Components of the ras signaling pathway contribute to activation of cellular p53. In MCF-7 cells, p38 kinase activated p53 more effectively than other members of the ras pathway. p53 and p38 kinase exist in the same physical complex, and co-expression
Knockdown of hnRNPK leads to increased DNA damage after irradiation and reduces survival of tumor cells
Wiesmann N, et al.
Carcinogenesis, 38(3), 321-328 (2017)
Hui Miao et al.
PLoS genetics, 15(5), e1008144-e1008144 (2019-05-16)
Long noncoding RNAs (lncRNAs) participate in various biological processes such as apoptosis. The function of lncRNAs is closely correlated with their localization within the cell. While regulatory potential of many lncRNAs has been revealed at specific subcellular location, the biological



Global Trade Item Number

SKUGTIN
P6874-200UL04061837868399

Questions

Reviews

No rating value

Active Filters