Skip to Content
Merck

Skip To

P4139

L-α-Phosphatidylcholine, hydrogenated

Type I-EH, ≥99%

Synonym(s):

1,2-Diacyl-sn-glycero-3-phosphocholine, L-α-Lecithin, PC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25
EC Number:
232-307-2
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


type

Type I-EH

Quality Segment

assay

≥99%

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1

InChI key

JLPULHDHAOZNQI-ZTIMHPMXSA-N

Preparation Note

Highly purified egg lecithin hydrogenated in the presence of a palladium catalyst.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P2772P427961755
functional group

phospholipid

functional group

phospholipid

functional group

phospholipid

functional group

phospholipid

assay

≥99%

assay

≥99%

assay

≥99% (TLC)

assay

-

form

powder

form

solution

form

solution

form

solid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

ambient

shipped in

-

shipped in

-

shipped in

ambient

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
P4139-1G04061826686201
P4139-100MG04061832796642

Questions

  1. What is its source? From soybeans or sunflowers?

    1 answer
    1. The raw material used to produce this product is chicken egg yolk.

      Helpful?

Reviews

No rating value

Active Filters