Skip to Content
Merck

Skip To

P1504

TWEEN® 40

viscous liquid

Synonym(s):

Polyoxyethylenesorbitan monopalmitate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12161900
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

non-ionic

Quality Segment

assay

~90.0% (GC)

form

viscous liquid

mol wt

estimated mol wt 1277

composition

Palmitic acid, ~90% (balance primarily stearic acid)

technique(s)

DNA extraction: suitable, RNA extraction: suitable, cell culture | mammalian: suitable, protein purification: suitable

CMC

0.027 (20-25°C)

HLB

15.6

SMILES string

O1C(C(C(C1)OCCO)OCCO)C(OCCO)COCCOC(=O)CCCCCCCCCCC

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

General description

Polyoxyethylenesorbitan monopalmitate, or TWEEN 40, is a non-ionic detergent.

Application

Non-ionic detergent used for cell lysis, nuclei isolation and cell fractionation.
Polyoxyethylenesorbitan monopalmitate, or TWEEN 40, has been used in a study to assess the determination of iodide in urine based on a chemiluminescence system. It has also been used in a study to investigate the delivery of astaxanthin and canthaxanthin to HepG2 cells. [1]

Legal Information

TWEEN is a registered trademark of Croda International PLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.22185STS0201655204
technique(s)

DNA extraction: suitable, cell culture | mammalian: suitable, RNA extraction: suitable, protein purification: suitable

technique(s)

-

technique(s)

-

technique(s)

-

form

viscous liquid

form

liquid

form

viscous liquid

form

liquid

assay

~90.0% (GC)

assay

-

assay

~90.0% (GC)

assay

-

CMC

0.027 (20-25°C)

CMC

-

CMC

0.027 (20-25°C)

CMC

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

mol wt

estimated mol wt 1277

mol wt

-

mol wt

estimated mol wt 1277

mol wt

~1228


Still not finding the right product?

Explore all of our products under TWEEN® 40


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Christine Boesch-Saadatmandi et al.
Cytotechnology, 63(1), 89-97 (2010-12-15)
The present investigation aimed to compare fetal calf serum (FCS) and Tween 40 with the commonly employed tetrahydrofuran (THF) with respect to cytotoxicity, stability of the solubilized carotenoids, and uptake and accumulation of the xanthophylls astaxanthin (AX) and canthaxanthin (CX)
F Podlogar et al.
International journal of pharmaceutics, 276(1-2), 115-128 (2004-04-29)
Pharmaceutically usable microemulsion systems were prepared from water and isopropyl myristate with a constant amount of Tween 40 and Imwitor 308 at a mass ratio of 1. Their type and structure were examined by measuring density and surface tension, and
Mohammad Reza Lornejad-Schäfer et al.
European journal of nutrition, 46(2), 79-86 (2007-01-18)
Carotenoids lutein and zeaxanthin are proposed to protect ocular tissues from free-radical damage that can cause cataract and age-related macular degeneration (AMD). They accumulate selectively in the lens and macular region of the retina. Changes in the retinal pigment epithelium



Global Trade Item Number

SKUGTIN
P1504-500ML04061837602290
P1504-1GA04061834355144
P1504-100ML04061837602283

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters