Skip to Content
Merck

Skip To

P118

Phaclofen

solid

Synonym(s):

3-Amino-2-(4-chlorophenyl)propanephosphonic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C9H13ClNO3P
CAS Number:
Molecular Weight:
249.63
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

Quality Segment

impurities

≥97% (NMR)

color

white

solubility

0.1 M HCl: 13.3 mg/mL, methanol: 19.3 mg/mL, DMSO: insoluble

SMILES string

NCC(CP(O)(O)=O)c1ccc(Cl)cc1

InChI

1S/C9H13ClNO3P/c10-9-3-1-7(2-4-9)8(5-11)6-15(12,13)14/h1-4,8H,5-6,11H2,(H2,12,13,14)

InChI key

VSGNGLJPOGUDON-UHFFFAOYSA-N

Gene Information

General description

Phaclofen is a phosphonic acid derivative of baclofen.

Application

Phaclofen has been used to block GABAB receptor.

Biochem/physiol Actions

Phaclofen plays a vital role in identifying the physiological importance of central and peripheral bicuculline-insensitive receptors with which GABA and (−) baclofen interact. Phaclofen acts as a GABAB receptor antagonist. Phaclofen has an ability to reversibly block the late, bicuculline resistant, K+ dependent inhibitory postsynaptic potential (IPSP) logged in projection cells of the cat and rat dorsal lateral geniculate nucleus and in rat hippocampal CA1 pyramidal neurons.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A6566B0200050S166
Quality Level

100

Quality Level

200

Quality Level

-

Quality Level

200

form

solid

form

solid

form

-

form

solid

Gene Information

human ... GABBR1(2550), GABBR2(9568)

Gene Information

human ... GABBR1(2550), GABBR2(9568)

Gene Information

-

Gene Information

human ... GABBR1(2550), GABBR2(9568)
mouse ... GABBR1(54393), GABBR2(242425)
rat ... GABBR1(81657), GABBR2(83633)

impurities

≥97% (NMR)

impurities

-

impurities

-

impurities

-

color

white

color

white

color

-

color

white

solubility

0.1 M HCl: 13.3 mg/mL, DMSO: insoluble, methanol: 19.3 mg/mL

solubility

H2O: 1 mg/mL, 0.1 M HCl: 1.2 mg/mL, methanol: 1.4 mg/mL, DMSO: 28 mg/mL, 0.1 M NaOH: 9.5 mg/mL

solubility

-

solubility

0.1 M NaOH: 20 mg/mL


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
P118-5MG04061834354208
P118-25MG04061833007945
P118-100MG04061833007938

Questions

Reviews

No rating value

Active Filters