Skip to Content
Merck

Skip To

P0376

Phenolphthalein β-D-glucuronide sodium salt

β-glucuronidase substrate, chromogenic, ≥90% (TLC), powder

Synonym(s):

Phenolphthalein glucuronic acid, Phenolphthalein mono-β-glucosiduronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C26H21O10Na
CAS Number:
Molecular Weight:
516.43
NACRES:
NA.28
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
229-896-3
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Phenolphthalein β-D-glucuronide sodium salt, β-glucuronidase substrate

Quality Segment

assay

≥90% (TLC)

form

powder

solubility

H2O: soluble 5 mg/mL

storage temp.

−20°C

SMILES string

[Na+].O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C([O-])=O)Oc2ccc(cc2)C3(OC(=O)c4ccccc34)c5ccc(O)cc5

InChI

1S/C26H22O10.Na/c27-15-9-5-13(6-10-15)26(18-4-2-1-3-17(18)24(33)36-26)14-7-11-16(12-8-14)34-25-21(30)19(28)20(29)22(35-25)23(31)32;/h1-12,19-22,25,27-30H,(H,31,32);/q;+1/p-1/t19-,20-,21+,22-,25+,26?;/m0./s1

InChI key

GBLJULUMBNYFAK-PDFLYTMFSA-M

Biochem/physiol Actions

Phenolphthalein β-D-glucuronide sodium salt is a β glucuronidase substrate. It is biosynthesized from the urine of rabbits to which phenolphthalein has been administered.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P0501P9875N1627
assay

≥90% (TLC)

assay

≥97% (TLC)

assay

~95%

assay

≥98% (TLC)

solubility

H2O: soluble 5 mg/mL

solubility

H2O: soluble 100 mg/mL

solubility

water: 50 mg/mL, clear, colorless to faint yellow or tan

solubility

H2O: soluble 100 mg/mL, clear

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters