Skip to Content
Merck

Skip To

O1141

Olanzapine

≥98% (HPLC), 5-HT2 serotonin and D1/D2 dopamine receptor antagonist, powder

Synonym(s):

LY-170053, 2-Methyl-4-(4-methyl-1-piperazinyl)- 10H-thieno[2,3-b][1,5]benzodiazepine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H20N4S
CAS Number:
Molecular Weight:
312.43
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Beilstein/REAXYS Number:
7655141
Assay:
≥98% (HPLC)
Form:
powder
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Olanzapine, ≥98% (HPLC)

Quality Level

assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: >15 mg/mL

originator

Eli Lilly

storage temp.

2-8°C

SMILES string

CN1CCN(CC1)C2=Nc3ccccc3Nc4sc(C)cc24

InChI

1S/C17H20N4S/c1-12-11-13-16(21-9-7-20(2)8-10-21)18-14-5-3-4-6-15(14)19-17(13)22-12/h3-6,11,19H,7-10H2,1-2H3

InChI key

KVWDHTXUZHCGIO-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Olanzapine is a 5-HT2 serotonin and D1/D2 dopamine receptor antagonist.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP1060Y0001380PHR1825
form

powder

form

solid

form

-

form

-

assay

≥98% (HPLC)

assay

-

assay

-

assay

-

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

300

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-30°C

solubility

DMSO: >15 mg/mL

solubility

-

solubility

-

solubility

-

color

yellow

color

-

color

-

color

-


Still not finding the right product?

Explore all of our products under Olanzapine


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


N A Moore et al.
The Journal of pharmacology and experimental therapeutics, 262(2), 545-551 (1992-08-01)
Olanzapine (LY170053, 2-methyl-4-(4-methyl-1-piperazinyl)-10H-thieno[2,3-b][1,5] benzodiazepine) is a novel "atypical" antipsychotic agent with 5-hydroxytryptamine2.dopamine D1/D2 antagonist activity and anticholinergic properties. In behavioral studies, olanzapine (1.25-10 mg/kg, p.o.) antagonizes apomorphine-induced climbing behavior in mice, demonstrating that the compound possesses D1/D2 antagonist activity in
Heating under high-frequency inductive conditions: application to the continuous synthesis of the neurolepticum olanzapine (Zyprexa).
Jan Hartwig et al.
Angewandte Chemie (International ed. in English), 52(37), 9813-9817 (2013-07-31)
W Wolfgang Fleischhacker et al.
Biological psychiatry, 65(6), 510-517 (2008-11-07)
Few studies have directly compared the efficacy and tolerability of atypical agents. This multicenter, randomized, double-blind study compared the efficacy and tolerability of aripiprazole (n = 355) with olanzapine (n = 348) in patients with schizophrenia experiencing acute relapse. After



Global Trade Item Number

SKUGTIN
O1141-10MG04061834213123
O1141-50MG04061834213130

Questions

Reviews

No rating value

Active Filters