Skip to Content
Merck

Skip To

N6019

Neutralizing Buffer

for Northern and Southern blotting, powder

Synonym(s):

Neutralizing Buffer for Blotting Techniques

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.25
UNSPSC Code:
12161700
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Level

form

powder

technique(s)

Southern blotting: suitable

foreign activity

DNase, RNase, none detected

storage temp.

room temp

General description

Sigma′s neutralization buffer is suitable for Southern blotting procedures to neutralize the pH of the gel following depurination and denaturation treatment or before transferring it to membranes.

Application

Suitable for neutralizing the gel before transfer to membranes during Southern procedures.

Preparation Note

The Neutralization Buffer is composed of 1M Tris-HCl and 1.5M NaCl.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S814085639G7663
technique(s)

Southern blotting: suitable

technique(s)

-

technique(s)

-

technique(s)

-

grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

form

powder

form

powder blend

form

powder blend

form

powder blend

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

foreign activity

DNase, RNase, none detected

foreign activity

DNAse, none detected, Endonuclease, none detected, Exonuclease, none detected, NICKase, none detected, RNAse, none detected

foreign activity

-

foreign activity

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Sambrook, J., and Russell, D. W.
Molecular Cloning: A Laboratory Manual, 6-6 (2001)
A southern blot assay for detection of hepatitis B virus covalently closed circular DNA from cell cultures
Cai D, et al.
Methods in Molecular Biology, 151-161 (2013)
Ausubel, F.M., et al.
Short Protocols in Molecular Biology (1995)



Global Trade Item Number

SKUGTIN
N6019-5X1L04061834117377
N6019-1L04061834117360

Questions

Reviews

No rating value

Active Filters