Skip to Content
Merck

Skip To

N3020

Nuclear Fast Red Solution

Synonym(s):

Calcium Red Solution , Kernechtrot Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12171500
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Level

shelf life

Expiry date on the label

IVD

for in vitro diagnostic use

application(s)

hematology
histology

storage temp.

room temp

SMILES string

[Na+].Nc1c(O)c(c(O)c2C(=O)c3ccccc3C(=O)c12)S([O-])(=O)=O

InChI

1S/C14H9NO7S.Na/c15-9-7-8(12(18)14(13(9)19)23(20,21)22)11(17)6-4-2-1-3-5(6)10(7)16;/h1-4,18-19H,15H2,(H,20,21,22);/q;+1/p-1

InChI key

IFSXZLJQEKGQAF-UHFFFAOYSA-M

General description

Nuclear Fast Red is an anthraquinone dye, and isalso called as calcium red.

Application

Used as a red nuclear counterstain. Nuclear Fast Red solution has been used:
  • In the dual-beam (FIB (focused ion beam)/SEM (scanning electron microscopy)) analysis of intracellular MNPs (magnetic nanoparticles) distribution.[1]
  • As a counterstain in LacZ tissue histological analysis, and in extracellular matrix proteoglycan staining.
  • In situ hybridization.

Other Notes

Nuclear fast red 0.1% in 5% aluminum sulfate.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
N800260700N6264
application(s)

hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

hematology
histology

form

solution

form

powder

form

powder

form

solution

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

IVD

for in vitro diagnostic use

IVD

-

IVD

-

IVD

for in vitro diagnostic use

shelf life

Expiry date on the label

shelf life

-

shelf life

-

shelf life

Expiry date on the label

Quality Level

500

Quality Level

200

Quality Level

100

Quality Level

500


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Yanli Gu et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 43(5), 2001-2009 (2017-10-23)
It is well known that Plac1 is a placenta-specific gene; however, its spatiotemporal expression pattern and exact role at t h e mouse fetomaternal interface r e m a i n s unclear. In situ hybridization (ISH) was used to
Morphological Plasticity of Emerging Purkinje Cells in Response to Exogenous VEGF
Herrfurth L, et al.
Frontiers in Molecular Neuroscience, 10(10), 2-2 (2017)
Naomi D Chrispijn et al.
PloS one, 13(7), e0200316-e0200316 (2018-07-10)
Polycomb Group (PcG) genes are transcriptional repressors that are described to be important during development and differentiation. There is significant interest in PcGs proteins because of their role in stem cell biology and tumorigenesis. In this study we characterize the



Global Trade Item Number

SKUGTIN
N3020-100ML04061834115687

Questions

  1. looking for instruction to prepare a working solution of nuclear fast red stain using 60700. should Aluminum Sulfate be added? If so, can Aluminum Sulphate hydrate be used? need to use it as a counter stain for my iron staining.

    1 answer
    1. The Nuclear Fast Red solution can be prepared following a specific protocol with catalog number N8002It is recommended to use aluminum sulfate anhydrous over the various hydrated forms of aluminum sulfate. The hydrated forms of aluminum sulfate may contain 30 to 50% less aluminum sulfate per gram of solid, potentially leading to shorter solution stability. The prepared solution is suitable for iron staining and other special stain procedures that require a non-blue nuclear counterstain.

      Helpful?

Reviews

No rating value

Active Filters