Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Quality Segment
assay
~98%
form
powder
packaging
pkg of 1 mg (per vial)
solubility
10 mM NaOH: 0.1 mg/mL, clear, colorless
SMILES string
[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@H]2O[C@@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@H](O)[C@@H]2O
InChI
1S/C21H29N7O14P2.2Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;;/h1,3-4,7-8,10-11,13-16,20-21,29-32H,2,5-6H2,(H2,23,33)(H,34,35)(H,36,37)(H2,22,24,25);;/q;2*+1/p-2/t10-,11+,13-,14+,15-,16+,20-,21+;;/m0../s1
InChI key
QRGNQKGQENGQSE-QUWMEQBESA-L
Application
Biochem/physiol Actions
Preparation Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| assay ~98% | assay ~98% | assay ~98% | assay ~98% |
| solubility 10 mM NaOH: 0.1 mg/mL, clear, colorless | solubility - | solubility - | solubility 10 mM NaOH: soluble-1 mg/mL, clear, colorless |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| form powder | form powder | form solid | form powder |
| packaging pkg of 1 mg (per vial) | packaging pkg of 0.2 mg (per vial) | packaging pkg of 2 mg (per vial) | packaging pkg of 10 mg (per vial) |
Still not finding the right product?
Explore all of our products under β-Nicotinamide adenine dinucleotide, reduced disodium salt
Storage Class
11 - Combustible Solids
wgk
WGK 3
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| N0786-1VL | 04061837143465 |
| N0786-15VL | 04061834093091 |
