Skip to Content
Merck

Skip To

MSRT2

SigmaProt Intact Protein LC-MS Standard

Proteomics Retention Time Standard

Synonym(s):

Intact Protein LC-MS Standard, SigmaProtein Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
23201100
EC Number:
220-163-3
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

ready-to-use solution

storage temp.

−20°C

Application

The RP-LC-MS Protein Standard is a LC-MS platform standard that is intended to test LC-MS system suitability and intact protein mass measurement. The proteins in MSRT2 are chosen to represent an elution profile for complex proteomic samples. The proteins have been selected based on UV and electrospray response, and are readily observed by both modes of detection. After reconstitution, MSRT2 is an injection-ready standard. MSRT2 can be utilized in studies such as in top-down proteomics.

Other Notes

MSRT2 contains these 9 proteins:
  • Ribonuclease B
  • Insulin
  • Lysozyme
  • Transferrin
  • Bovine serum albumin (BSA)
  • Trypsin inhibitor
  • β-lactoglobulin A
  • Carbonic anhydrase
  • Lactate dehydrogenase

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P8119P5369P5494
storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

ready-to-use solution

form

liquid

form

-

form

-




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. There is no information about the proteins' concentration, could you please add it?

    1 answer
    1. Each vial contains 100 ug each of the 9 constituent proteins, in lyophilized form. The recommended solvent is 0.1% trifluoroacetic acid (TFA) in water. Please see the link below to review additional information available in the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/421/524/msrt2bul.pdf

      Helpful?

Reviews

No rating value

Active Filters