Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
quality
Phosphopeptide Standard for MS
Quality Segment
analyte chemical class(es)
amino acids, peptides, proteins
packaging
pkg of 200 pmol total phosphopeptides
technique(s)
HPLC: suitable, LC/MS: suitable
application(s)
food and beverages
format
multi-component solution
storage temp.
−20°C
General description
Each of the three phosphopeptides mixes are available in their naturally occurring isotopic abundances (light) or as stable isotope enriched versions (heavy), making the set of products highly amenable to quantitative analyses, allowing users to compare recovery between workflows or techniques.
- Naturally occurring peptide sequences
- Broad range of peptide characteristics
- Complementary product designs
- Available in light and heavy versions
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| format multi-component solution | format multi-component solution | format multi-component solution | format multi-component solution |
| technique(s) HPLC: suitable, LC/MS: suitable | technique(s) HPLC: suitable, LC/MS: suitable | technique(s) HPLC: suitable, LC/MS: suitable | technique(s) HPLC: suitable, LC/MS: suitable |
| packaging pkg of 200 pmol total phosphopeptides | packaging pkg of 200 pmol total phosphopeptides | packaging pkg of 200 pmol total phosphopeptides | packaging pkg of 200 pmol total phosphopeptides |
| analyte chemical class(es) amino acids, peptides, proteins | analyte chemical class(es) amino acids, peptides, proteins | analyte chemical class(es) amino acids, peptides, proteins | analyte chemical class(es) amino acids, peptides, proteins |
| application(s) food and beverages | application(s) food and beverages | application(s) food and beverages | application(s) food and beverages |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



