Skip to Content
Merck

Skip To

M8250

MES hydrate

≥99.5% (titration)

Synonym(s):

2-(N-Morpholino)ethanesulfonic acid hydrate, 4-Morpholineethanesulfonic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H13NO4S · xH2O
CAS Number:
Molecular Weight:
195.24 (anhydrous basis)
UNSPSC Code:
12161700
eCl@ss:
32129211
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Chemical/Synthestic)

Quality Level

assay

≥99.5% (titration)

form

crystalline powder

storage condition

dry at room temperature

technique(s)

cell culture | plant: suitable, immunofluorescence: suitable

impurities

≤5 ppm (Total: Ag, As, Bi, Cd, Cu, Hg, Mo, Pb, Sb, Sn)

color

white

useful pH range

5.5-6.7

pKa 

6.1

solubility

water: 0.25 g/mL, clear, colorless

suitability

suitable for electrophoresis

application(s)

agriculture
clinical research
diagnostic assay manufacturing
life science and biopharma

storage temp.

room temp

SMILES string

O.OS(=O)(=O)CCN1CCOCC1

InChI

1S/C6H13NO4S.H2O/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);1H2

InChI key

MIIIXQJBDGSIKL-UHFFFAOYSA-N

General description

MES hydrate, or 4-morpholineethanesulfonic acid hydrate, is a zwitterionic and morpholinic buffer renowned as one of the "Good′s" buffer for biological and biochemical applications. Its outstanding buffering capacity in the pH range of 5.5 to 6.7 makes it versatile for a wide array of research applications. Notably, MES Buffer minimizes interaction with common metals used in environmental and biological studies, preventing interference with experimental outcomes.

With excellent water solubility and virtual insolubility in lipids, MES Buffer is impermeable to membranes, ensuring efficacy in cellular environments. Its applications span maintaining stability in cell culture media, protein-based buffer formulations, and electrophoresis running buffers. MES Buffer may also find usage in purifying antibodies, peptides, proteins, blood components, and growth factors. MES Buffer stands out as a better alternative to potentially toxic agents like cacodylate and non-zwitterionic buffers such as citrate and malate. MES Buffer proves valuable for adjusting growth media pH, stabilizing enzymatic solutions, and serving as a component of PAGE running buffer in diverse experimental settings.

Application

MES hydrate has been used:
  • in the MS (Murashige and Skoog) media for growth of Arabidopsis seedlings.[1]
  • in the cytoskeleton buffer during immunofluorescence study.
  • as a wash buffer to wash the paramagnetic beads and resuspend them for binding mannose-BSA
  • as a component of growth medium

Features and Benefits

  • Effective Buffering from pH 5.5-6.7 with a pKa of 6.1 (25 °C)
  • Highly soluble in water
  • Minimal metal ion binding
  • Stable in a wide pH range
  • Low UV absorptivity and Minimal reactivity

Preparation Note

Solutions are stable at 2-8°C for months. Sterilize by filtration through 0.2uM filters. Autoclaving is not recommended for any sulfonic acid buffer. If buffers must be nuclease-free, treat the water first, and then add the buffer after autoclaving. When MES solutions are autoclaved, they turn yellow (although pH does not change measurably). The identity of the yellow breakdown product is unknown.
A buffer using MES can be prepared by titrating with NaOH to the desired pH. Alternatively, stock solutions of MES and MES sodium salt can be mixed to attain the desired pH. Standard mixing tables using stock solutions to prepare buffers of a given pH have been published. MES is not recommended for buffering at pH 7.4; other buffers should be considered.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M528769890RDD030
assay

≥99.5% (titration)

assay

≥99.5% (titration)

assay

≥99.5% (T)

assay

≥99.0%

technique(s)

cell culture | plant: suitable, immunofluorescence: suitable

technique(s)

-

technique(s)

-

technique(s)

-

Quality Level

400

Quality Level

400

Quality Level

200

Quality Level

100

application(s)

agriculture
clinical research
diagnostic assay manufacturing
life science and biopharma

application(s)

cell analysis
life science and biopharma
sample preparation

application(s)

-

application(s)

-

form

crystalline powder

form

crystalline powder

form

powder or crystals

form

-

solubility

water: 0.25 g/mL, clear, colorless

solubility

H2O: 0.5 M, clear, colorless, water: 335.3 g/L at 20 °C

solubility

H2O: 0.5 M at 20 °C, clear, colorless

solubility

-


Still not finding the right product?

Explore all of our products under MES hydrate


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

MES Specification Comparison


Proteomic and phosphoproteomic analyses of chromatin-associated proteins from Arabidopsis thaliana.
Bigeard J et al.
Proteomics, 14, 2141-2141 (2014)
Data for Biochemical Research, Dawson, R.M.C. et al, 410-410 (1987)
"(Un) suitability of the use of pH buffers in biological, biochemical and environmental studies and their interaction with metal ions-a review.
Ferreira, Carlos MH, et al.
Royal Society of Chemistry Advances, 5 (39), 30989-31003 (2015)



Global Trade Item Number

SKUGTIN
M8250-1KG04061835265497
M8250-250G04061835265503
M8250-25KG04061832549545
M8250-25G04061834062967
M8250-50KG04061832549552
M8250-100G04061835265480

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters