Skip to Content
Merck

Skip To

M7167

M2 medium

With HEPES, without penicillin and streptomycin, liquid, sterile-filtered, suitable for mouse embryo cell culture

Synonym(s):

Cell Culture M2, Cell Culture Medium, M2 Medium Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

sterile-filtered

form

liquid

storage condition

protect from light

technique(s)

cell culture | embryo: suitable

impurities

endotoxin, tested

components

HEPES: 5.42726 g/L
NaHCO3: 0.35 g/L
glucose: 1.0 g/L (Dextro)
phenol red: 0.0106 g/L
sodium pyruvate: 0.0363 g/L

shipped in

ambient

storage temp.

2-8°C

Application

Recommended for manipulation of mouse embryos at ambient temperature. M2 and M16 Medium are common media for in vitro culture of preimplantation stage embryos. It is a modified Krebs-Ringer bicarbonate solution, which is very similar to Whitten′s Medium. M16 contains pyruvate and lactate as energy sources since preimplantation embryos do not utilize glucose efficiently.

M2 Medium is a further modification of M16 that substitutes HEPES buffer in place of some of the bicarbonate. M2 is used for collecting and handling embryos for prolonged periods outside a CO2 incubator.

Preparation Note

Supplement with 0.06 g/L potassium penicillin-G and 0.05 g/L streptomycin sulfate.

Analysis Note

Presence of particulates in solution will not affect product′s performance.
This product is tested for its ability to support the development of one-cell mouse embryos to expanded blastocysts. Minimum requirement is 80% development to blastocyst.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M729214660C14610C
form

liquid

form

liquid

form

liquid

form

liquid

technique(s)

cell culture | embryo: suitable

technique(s)

cell culture | embryo: suitable

technique(s)

cell culture | hybridoma: suitable, cell culture | mammalian: suitable

technique(s)

cell culture | hybridoma: suitable, cell culture | mammalian: suitable

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

components

HEPES: 5.42726 g/L
NaHCO3: 0.35 g/L
glucose: 1.0 g/L (Dextro)
phenol red: 0.0106 g/L
sodium pyruvate: 0.0363 g/L

components

NaHCO3: 2.101 g/L
phenol red: 0.0106 g/L
sodium pyruvate: 0.0363 g/L
glucose: 1.0 g/L (Dextro)

components

L-glutamine: no
glucose: 4.0 g/L
HEPES: 7.5 mM
NaHCO3: 1.05 g/L

components

glucose: 3.45 g/L
NaHCO3: 1.60 g/L
L-glutamine: 0.333 g/L
HEPES: 7.5 mM

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient


Still not finding the right product?

Explore all of our products under M2 medium


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Maryse Lessard et al.
Scientific reports, 9(1), 13829-13829 (2019-09-27)
The paternal environment is thought to influence sperm quality and future progeny may also be impacted. We hypothesized that prenatal exposure to environmentally-relevant contaminants impairs male reproduction, altering embryo gene expression over multiple generations. Folic acid (FA) can improve sperm
Hongzheng Sun et al.
Biology of reproduction, 100(3), 601-617 (2018-09-29)
Methionine adenosyltransferase II (MAT2A) is essential to the synthesis of S-adenosylmethionine, a major methyl donor, from L-methionine and ATP. Upon fertilization, zygotic genome activation (ZGA) marks the period that transforms the genome from transcriptional quiescence to robust transcriptional activity. During
Ludwig Stenz et al.
PloS one, 14(6), e0208371-e0208371 (2019-06-11)
Di(2-ethylhexyl)phthalate (DEHP) interferes with sex hormones signaling pathways (SHP). C57BL/6J mice prenatally exposed to 300 mg/kg/day DEHP develop a testicular dysgenesis syndrome (TDS) at adulthood, but similarly-exposed FVB/N mice are not affected. Here we aim to understand the reasons behind



Global Trade Item Number

SKUGTIN
M7167-50ML04061834061687
M7167-100ML04061838259820

Questions

Reviews

No rating value

Active Filters