Skip to Content
Merck

Skip To

M3183

MOPS

≥99.5% (titration), crystalline powder, BioPerformance Certified, suitable for cell culture

Synonym(s):

3-(N-Morpholino)propanesulfonic acid, 4-Morpholinepropanesulfonic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C7H15NO4S
CAS Number:
Molecular Weight:
209.26
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12161700
EC Number:
214-478-5
MDL number:
Beilstein/REAXYS Number:
1106776
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)

grade

BioPerformance Certified

Quality Segment

assay

≥99.5% (titration)

form

crystalline powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin and TAMC, tested

pH

2.5-4 (25 °C, 209 g/L)

useful pH range

6.5-7.9

pKa (25 °C)

7.2

cation traces

Fe: ≤5 ppm

λ

33 % in H2O

application(s)

diagnostic assay manufacturing

foreign activity

DNase, RNase, protease, none detected

SMILES string

OS(=O)(=O)CCCN1CCOCC1

InChI

1S/C7H15NO4S/c9-13(10,11)7-1-2-8-3-5-12-6-4-8/h1-7H2,(H,9,10,11)

InChI key

DVLFYONBTKHTER-UHFFFAOYSA-N

Application


  • A production platform for disulfide-bonded peptides in the periplasm of Escherichia coli.: This research developed a novel platform for producing disulfide-bonded peptides in E. coli, using MOPS buffer to maintain a stable pH environment essential for protein folding and stability in the periplasmic space. The platform could significantly enhance the production of therapeutically relevant peptides (Gibisch et al., 2024).


Packaging

M3183-1KT:
Each kit contains 3 x 100G samples, each sample from a uniquely manufactured lot.

Other Notes

Easily compare specifications for MOPS products with the MOPS specification table.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M5162M5162M1254
technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable, electrophoresis: suitable

technique(s)

cell culture | mammalian: suitable, electrophoresis: suitable

technique(s)

affinity chromatography: suitable, electrophoresis: suitable

assay

≥99.5% (titration)

assay

≥99.5% (titration)

assay

≥99.5% (titration)

assay

≥99.5% (titration)

grade

BioPerformance Certified

grade

reagent grade

grade

reagent grade

grade

-

Quality Level

300

Quality Level

400

Quality Level

400

Quality Level

200

form

crystalline powder

form

powder

form

powder

form

crystalline powder

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

advanced drug delivery
clinical research
diagnostic assay manufacturing
life science and biopharma

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable, electrophoresis: suitable

technique(s)

cell culture | mammalian: suitable, electrophoresis: suitable

technique(s)

affinity chromatography: suitable, electrophoresis: suitable

assay

≥99.5% (titration)

assay

≥99.5% (titration)

assay

≥99.5% (titration)

assay

≥99.5% (titration)

grade

BioPerformance Certified

grade

reagent grade

grade

reagent grade

grade

-

Quality Level

300

Quality Level

400

Quality Level

400

Quality Level

200

form

crystalline powder

form

powder

form

powder

form

crystalline powder

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

advanced drug delivery
clinical research
diagnostic assay manufacturing
life science and biopharma


Still not finding the right product?

Explore all of our products under MOPS


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters