Skip to Content
Merck

Skip To

L3755

Lipopolysaccharides from Escherichia coli O26:B6

purified by trichloroacetic acid extraction

Lipopolysaccharides from Escherichia coli O26:B6

Synonym(s):

LPS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
EC Number:
297-473-0
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Escherichia coli (O26:B6)

Quality Segment

form

lyophilized powder

purified by

trichloroacetic acid extraction

impurities

1-10% Protein (Lowry)

color

white to yellow

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

General description

This product is TCA extracted from E. coli serotype O26:B6. The source strain is ATCC 12795. LPS from serotype 026:B6 is the only LPS from E. coli which exhibits short chain-length behavior on SDS-PAGE. The short chain-length of this LPS is closer to that of the mutant rough strain LPS.

Application

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).

Biochem/physiol Actions

Lipopolysaccharides (LPS) are localized in the outer layer of the membrane and are, in noncapsulated strains, exposed on the cell surface. They contribute to the integrity of the outer membrane, and protect the cell against the action of bile salts and lipophilic antibiotics.

Preparation Note

The product is soluble in water (5 mg/ml) or cell culture medium (1 mg/ml) yielding a hazy, faint yellow solution. A more concentrated, though still hazy, solution (20 mg/ml) has been achieved in aqueous saline after vortexing and warming to 70-80 oC. Lipopolysaccharides are molecules that form micelles in every solvent. Hazy solutions are observed in water and phosphate buffered saline. Organic solvents do not give clearer solutions. Methanol yields a turbid suspension with floaters, while water yields a homogeneously hazy solution.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L4005L2762L8274
biological source

Escherichia coli (O26:B6)

biological source

Escherichia coli (O55:B5)

biological source

Escherichia coli (O26:B6)

biological source

Escherichia coli (O26:B6)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

solubility

water: soluble

solubility

water: soluble

solubility

water: 4.90-5.10 mg/mL, cloudy to hazy, colorless to faintly yellow

solubility

water: soluble

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

impurities

1-10% Protein (Lowry)

impurities

1-10% Protein (Lowry)

impurities

<5% Protein

impurities

<3% Protein (Lowry)


Still not finding the right product?


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
L3755-100MG04061833952290
L3755-500MG04061833952306

Questions

Reviews

Active Filters

  1. Israel
    • Review 1
    • Votes 0
    5 out of 5 stars.

    LPS L3755

    Works excellent in the animal model.................

    Helpful?