Skip to Content
Merck

Skip To

L0288

Lipid Mixture 1, Chemically Defined

liquid, sterile-filtered, BioReagent, suitable for cell culture

Synonym(s):

lipid mixture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.75
UNSPSC Code:
12352211
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

sterile-filtered

Quality Level

product line

BioReagent

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

storage temp.

2-8°C

General description

Lipid mixture 1, Chemically defined is recommended for use in cell culture at 1 to 10 mL per liter of medium.

Application

Lipid Mixture 1, Chemically Defined has been used:

  • as a component of bovine serum albumin (BSA) containing Bracket-Oliphant (BO) medium for the resuspension of sperm pellet[1]
  • as a supplement in spheroid media for culturing primary human subcutaneous stromal vascular fraction cells[2]
  • to prepare a cell steatosis model for culturing of mouse and human hepatocytes[3]

Other Notes

Contains non-animal derived fatty acids (2 μg/ml arachidonic and 10 μg/ml each linoleic, linolenic, myristic, oleic, palmitic and stearic), 0.22 mg/ml cholesterol from New Zealand sheep′s wool, 2.2 mg/ml Tween-80, 70 μg/ml tocopherol acetate and 100 mg/ml Pluronic F-68 solubilized in cell culture water.
The final product contains 10% ethanol by volume.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L5146D0547D0697
technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | insect: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

form

liquid

form

liquid

form

powder

form

liquid

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

500

Quality Level

500

Quality Level

500

Quality Level

400

sterility

sterile-filtered

sterility

sterile-filtered

sterility

-

sterility

sterile-filtered

product line

BioReagent

product line

BioReagent

product line

-

product line

-


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Anna Ioannidou et al.
The Journal of physiology (2021-08-14)
Adipocyte enlargement is a key feature of obesity and associated with insulin resistance and metabolic disease The cause and consequences of adipocyte enlargement have remained hard to study in vitro due to a lack of human cell models with representative
Juan Zhong et al.
International immunopharmacology, 59, 197-208 (2018-04-16)
The anti-inflammatory, immunomodulatory, and anticancer effects of micheliolide (MCL) isolated from Michelia champaca were previously reported, but its role and underlying mechanisms in relieving liver steatosis remain unclear. Herein, we investigated the effects of MCL on hepatic steatosis using a
Marc P Hübner et al.
PLoS neglected tropical diseases, 14(7), e0008427-e0008427 (2020-07-07)
A major impediment to eliminate lymphatic filariasis and onchocerciasis is the lack of effective short-course macrofilaricidal drugs or regimens that are proven to be safe for both infections. In this study we tested oxfendazole, an anthelmintic shown to be well



Global Trade Item Number

SKUGTIN
L0288-100ML04061833945650

Questions

1–4 of 4 Questions  
  1. does the L0288 have BSA in it or not? as the description is somewhat confusing?

    1 answer
    1. This product does not contain BSA (Bovine Serum Albumin) . In addition to the listed fatty acids, this product also contains Tween-80 at 70 μg/ml, tocopherol acetate at 100 mg/ml, Pluronic F-68, and 10% ethanol by volume.

      Helpful?

  2. 組成を教えて頂けませんでしょうか?

    1 answer
    1. As listed in the 'DESCRIPTION' section of the product page, under 'Other Notes', this product contains non-animal derived fatty acids (2 μg/ml arachidonic and 10 μg/ml each linoleic, linolenic, myristic, oleic, palmitic and stearic), 0.22 mg/ml cholesterol from New Zealand sheep's wool, 2.2 mg/ml Tween-80, 70 μg/ml tocopherol acetate and 100 mg/ml Pluronic F-68 solubilized in cell culture water.

      Helpful?

  3. Lipid Mixture 1, Chemically Defined(L0288) 제품 설명에서 non-animal source라고 되어 있는데 "0.22 mg/ml cholesterol from New Zealand sheep′s wool" 라는 문구가 있습니다. 이를 보면 콜레스테롤의 경우 양모에서 추출한 것 같은데, 혹시 이 성분은 animal source가 맞는건지 아니면 non-animal source인 건지 궁금합니다.

    1 answer
    1. L0288 is not derived from animal origin except for 2003140 cholesterol as stated in the sample CoA on the product site.

      Helpful?

  4. 안녕하세요. 위 제품에는 BSA (Bovine serum albumin)이 구성요소에 포함되어 있는것일까요? 아니면 BSA구성요소의 일부가 포함되어 있는 것일까요? BSA대체제로 사용할 수 있는 것일까요?

    1 answer
    1. As listed in the DESCRIPTION section, this product has been cited as a component of bovine serum albumin in a Bracket-Oliphant (BO) medium for the resuspension of sperm pellets. However, this product is typically used as a medium supplement in mammalian cell culture. While there are fatty acids present in BSA, this product alone is not a suitable replacement for bovine serum albumin. Other supplements would be required. The supplement profile would depend on the type of cell being cultured.

      To discuss further, please navigate to the link https://www.sigmaaldrich.com/techservice, click on "Product Technical Inquiries" under the Products Section, and respond with all the required information so that a member of the Technical Service team can reach out to assist further.

      For additional information, please see the link below to review the product datasheet for this supplement product as well as others:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/309/806/i3146pis.pdf

      Helpful?

Reviews

No rating value

Active Filters