Skip to Content
Merck

Skip To

K0640

Anti-Potassium Channel Kv11.1 (HERG) Extracellular−FITC antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-KCNH2, Anti-Voltage gated K+ channel subfamily H member 2, Anti-ether-a-go-go-related channel 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
FITC conjugate
Clone:
polyclonal
Application:
FACS, ICC
Citations:
4
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

flow cytometry: 2-10 μg/mL using 1x106 human live cells, immunocytochemistry: 1:25 using intact live HEK-KV11.1 transfected cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNH2(3757)

General description

Anti-Potassium Channel KV11.1 (HERG)- Extracellular-FITC (Voltage gated K+ channel subfamily H member 2, KCNH2, ether-a-go-go-related channel 1) is produced in rabbit using as immunogen the peptide AFLLKETEEGPPATEC corresponding to residues 430-445 of human KV11.1 (HERG). Anti-KV11.1 (HERG) antibody is directed against an extracellular epitope located between the S1 and S2 domains. The antibody is affinity purified on immobilized antigen and labeled with fluorescein isothiocyanate (FITC).
The gene Kv11.1, also referred to as KCNH2 (potassium voltage-gated channel subfamily H member 2), is mapped to human chromosome 7. The encoded voltage-activated potassium channel is a member of the ether-a-go-go (EAG) family.

Immunogen

peptide AFLLKETEEGPPATEC corresponding to residues 430-445 of human KV11.1 (HERG).

Application

Flow cytometry: a recommended working concentration of 2-10 μg per 1x106cells was determined using intact live human cells.

Biochem/physiol Actions

The gene Kv11.1, also referred to as HERG (human ether-a-go-go related gene), encodes the α-subunit of the rapid component of the delayed rectifier potassium channel. The third repolarization of the action potential of human cardiomyocytes is carried out by this protein. Mutations in this gene have been associated with type II hereditary long QT syndrome (LQT2). This disease is characterized by prolongation of the QT interval, abnormal T wave, torsade de pointes, syncope and sudden death caused by cardiac arrest.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide.

Preparation Note

Reconstitute the lyophilized vial with 50 μL or 200 μL deionized water, depending on package size. Further dilutions should be made using a carrier protein such as BSA (1-3%).

Other Notes

Lyophilized powder can be stored intact at room temperature for several weeks. For extended storage, it should be stored at -20 °C or below. The reconstituted solution can be stored at 2-8 °C for up to 2 weeks. For longer storage, freeze in working aliquots. Avoid repeated freezing and thawing. Storage in “frost-free” freezers is not recommended. Centrifuge before use. Working dilution samples should be discarded if not used within 12 hours. The antibody is stable for at least 12 months when stored appropriately.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P0749P4247P4497
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

FITC conjugate

conjugate

unconjugated

conjugate

FITC conjugate

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

species reactivity

human

species reactivity

human

species reactivity

rat, human, mouse

species reactivity

human, rat


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions






Global Trade Item Number

SKUGTIN
K0640-50UL04061837813320

Questions

1–2 of 2 Questions  
  1. What is the concentration of Product K0640, Anti-Potassium Channel Kv11.1 (HERG) Extracellular - FITC antibody produced in rabbit after reconstitution with 50 ul of water?

    1 answer
    1. Product K0640, Anti-Potassium Channel Kv11.1 (HERG) Extracellular - FITC antibody produced in rabbit will have a concentration of 1 mg/ml when reconstituted with 50 ul of water.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters