Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.56
UNSPSC Code:
12352203
Pricing and availability is not currently available.
antibody product type
primary antibodies
Quality Segment
clone
monoclonal
analyte chemical class(es)
proteins (hemagglutinin)
technique(s)
immunoprecipitation (IP): suitable
storage temp.
2-8°C
General description
Anti-hemagglutinin (HA) Immunoprecipitation Kit is designed to recover maximum amounts of HA-tagged proteins in the immunoprecipitates. The process is carried out in mini-spin columns instead of microcentrifuge tubes to ensure easy washing of antigen-antibody-bound beads with minimal variability.
Application
Anti- hemagglutinin (HA) Immunoprecipitation Kit has been used for immunoprecipitation. It has also been used in in vitro acetylation assay from human embryonic kidney cell lines.
Features and Benefits
- Anti- hemagglutinin (HA) coupled beads enable high specific binding to HA-tagged fusion proteins
- Coupling of Anti-HA antibodies to the beads eliminates other preliminary and calibration steps
- Mini-spin columns enable the minimal loss of anti-HA-bound beads during washing
- The CelLyticTM M cell lysis reagent provides a high yield, rapid and efficient mammalian cell lysis
- Ready-to-use cell lysis reagent and 10X concentrated immunoprecipitation buffer
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) immunoprecipitation (IP): suitable | technique(s) immunoprecipitation (IP): suitable | technique(s) - | technique(s) - |
| Quality Level 200 | Quality Level 300 | Quality Level 200 | Quality Level 200 |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. - | storage temp. −20°C |
| clone monoclonal | clone - | clone - | clone - |
| analyte chemical class(es) proteins (hemagglutinin) | analyte chemical class(es) proteins (FLAG® Affinity Gels) | analyte chemical class(es) - | analyte chemical class(es) - |
| antibody product type primary antibodies | antibody product type - | antibody product type - | antibody product type - |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



