Skip to Content
Merck

Skip To

I2142

KN-62

≥95%, powder

Synonym(s):

(S)-5-Isoquinolinesulfonic acid 4-[2-[(5-isoquinolinylsulfonyl)methylamino]-3-oxo-3-(4-phenyl-1-piperazinyl)propyl]phenyl ester, 1-[N,O-bis(5-Isoquinolinesulfonyl)-N-methyl-L-tyrosyl]-4-phenylpiperazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C38H35N5O6S2
CAS Number:
Molecular Weight:
721.84
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥95%
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥95%

form

powder

color

yellow

mp

130 °C

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 0.93 mg/mL, methanol: 10 mg/mL, DMSO: 12 mg/mL, chloroform: 20 mg/mL

storage temp.

−20°C

SMILES string

O=S(OC1=CC=C(C[C@@H](C(N2CCN(C3=CC=CC=C3)CC2)=O)N(C)S(C4=CC=CC5=C4C=CN=C5)(=O)=O)C=C1)(C6=CC=CC7=C6C=CN=C7)=O

InChI

1S/C38H35N5O6S2/c1-41(50(45,46)36-11-5-7-29-26-39-19-17-33(29)36)35(38(44)43-23-21-42(22-24-43)31-9-3-2-4-10-31)25-28-13-15-32(16-14-28)49-51(47,48)37-12-6-8-30-27-40-20-18-34(30)37/h2-20,26-27,35H,21-25H2,1H3/t35-/m0/s1

InChI key

RJVLFQBBRSMWHX-DHUJRADRSA-N

Gene Information

human ... P2RX7(5027)

Biochem/physiol Actions

Selectively inhibits rat brain Ca2+/calmodulin-dependent protein kinase II.

Features and Benefits

This compound is featured on the Ca/CaMKs and P2 Receptors: P2X Ion Channel Family pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2384S8010N2790
assay

≥95%

assay

≥98% (HPLC)

assay

-

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

200

Quality Level

-

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 0.93 mg/mL, DMSO: 12 mg/mL, methanol: 10 mg/mL, chloroform: 20 mg/mL

solubility

DMSO: 2 mg/mL, clear

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 8.0 mg/mL, 0.1 M HCl: soluble, H2O: slightly soluble, ethanol: soluble

solubility

DMSO: >15 mg/mL

color

yellow

color

white to beige

color

white

color

yellow


Still not finding the right product?

Explore all of our products under KN-62


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters