Skip to Content
Merck

Skip To

H2500

Hemoglobin from bovine blood

lyophilized powder

Synonym(s):

Bovine hemoglobin, Hb, Methemoglobin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
eCl@ss:
42030116
NACRES:
NA.61
MDL number:
Form:
lyophilized powder
Biological source:
bovine blood
Mol wt:
Mr ~64500
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine blood

Quality Level

form

lyophilized powder

mol wt

Mr ~64500

technique(s)

MALDI-TOF: suitable

UniProt accession no.

storage temp.

2-8°C

General description

Hemoglobin is the major component of red blood cells, and is responsible for their red color. Its normal concentration in erythrocytes is 34%. Hemoglobin is a globular protein with α and β chains with each 141 and 146 amino acids, respectively. It exists as a tetramer with each monomer having heterocyclic porphyrin ring with iron constituting the heme.

Application

The solubility of α-elastin has been applied to construction of elastin-mimetic biomaterials.
Hemoglobin from bovine blood has been used in:
  • standard curve generation for the quantification intraparenchymal hemorrhage and parenchymal hemorrhage in spinal cord homogenate using Drabkin′s assay, Quadrupole-Ion Mobility-Time-of-Flight mass spectrometery
  • the generation of molecularly imprinted polymers (MIPs) to mimic high molecular-weight polyethylene glycol (PEG) in crystallization studies

Biochem/physiol Actions

Oxygen transporter, NO scavenger
Oxygen transporter. The Fe2+/Fe3+ balance is a physiological indicator of blood oxygenation; deoxygenated hemoglobin accessorizes a feedback loop by reducing nitrite to NO, a vasodilator which enhances blood flow to oxygen-deprived tissues.
Hemoglobin is the most important respiratory protein of vertebrates by virtue of its ability to transport oxygen from the lungs to body tissues, and to facilitate the return transport of carbon dioxide. Bovine hemoglobin is used in the generation of hemoglobin-vesicles (HbV) for oxygen transport. It is thermally stable and displays high affinity to oxygen when compared to human hemoglobin. Bovine hemoglobin interacts with synthetic and azo dyes. Polymerized forms of bovine haemoglobin is recommended for treating autoimmune hemolytic anemia.

Disclaimer

Since native hemoglobin is readily oxidized in air, these preparations may be predominantly methemoglobin.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
H262508449H3760
biological source

bovine blood

biological source

bovine blood

biological source

bovine blood

biological source

bovine blood

technique(s)

MALDI-TOF: suitable

technique(s)

activity assay: suitable

technique(s)

-

technique(s)

-

mol wt

Mr ~64500

mol wt

Mr ~64500

mol wt

Mr ~64500

mol wt

Mr ~64500

form

lyophilized powder

form

substrate powder

form

powder

form

dried erythrocytes

UniProt accession no.

P01966, P02081, P06643, P02070, P06642

UniProt accession no.

P01966, P02070, P02081, P06642, P06643

UniProt accession no.

P01966, P02070, P02081, P06642, P06643

UniProt accession no.

P01966, P02070, P02081, P06642, P06643

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Hemoglobin from bovine blood


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Endogenous Interleukin-10 Deficiency Exacerbates Vascular Pathology in Traumatic Cervical Spinal Cord Injury
Badner A, et al.
Journal of Neurotrauma (2019)
Early Intravenous Infusion of Mesenchymal Stromal Cells Exerts a Tissue Source Age-Dependent Beneficial Effect on Neurovascular Integrity and Neurobehavioral Recovery After Traumatic Cervical Spinal Cord Injury
Vawda R, et al.
Stem Cells Translational Medicine (2019)
Extensive charge reduction and dissociation of intact protein complexes following electron transfer on a quadrupole-ion mobility-time-of-flight MS
Lermyte F, et al.
Journal of the American Society For Mass Spectrometry, 26(7), 1068-1076 (2015)



Global Trade Item Number

SKUGTIN
H2500-10G04061833630280
H2500-25G04061833792162
H2500-1G04061833792148
H2500-5G04061833792193

Questions

1–3 of 3 Questions  
  1. I want to create hemoglobin standards to use in a colorimetric assay. Can I create hemoglobin concentrations by weighing specific amounts of lyophilized hemoglobin powder and reconstituting with TS saline? Then process in assay after adding reagent?

    1 answer
    1. The solubility of this product has been tested in water at a concentration of 20 mg/mL, resulting in dark brown-red solutions.  Unfortunately, we have not tested this product in colorimetric assays and can provide no recommendations on this.  

      Helpful?

  2. Does the website specify if H2500 Hemoglobin from bovine blood is ferrohemoglobin or oxyhemoglobin?

    1 answer
    1. The website indicates that the product H2500 is mainly methemoglobin. A search of the website did not uncover any products marketed as either ferrohemoglobin or oxyhemoglobin.

      Helpful?

  3. What is the difference between H2500 and H2625 when making Chocolate agar with bovine hemoglobin? Although they share the same CAS and Specification Sheet, there is a significant difference in price between the two.

    1 answer
    1. Products H2500 and H2625 are not suitability tested for use in producing Chocolate Agar. The key differences between H2500 and H2625 lie in their purity and intended uses. H2500 is a purer product, suitable for MALDI-TOF, and associated with a companion product for HPLC analysis of hemoglobin variants. It undergoes two steps of ammonium sulfate precipitation, filtration, dialysis, and lyophilization. On the other hand, H2625 is not suitability tested for use as a protease substrate and lacks the ammonium sulfate precipitation step. These distinctions likely contribute to the significant price difference between the two products.

      Helpful?

Reviews

No rating value

Active Filters