Skip to Content
Merck

Skip To

GL0010

Golgi Isolation Kit

sufficient for 50 g (tissue)

Synonym(s):

Golgi Kit, Isolation Kit for Golgi

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

usage

sufficient for 50 g (tissue)

technique(s)

fractionation: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

The Golgi Isolation Kit provides a method for isolating Golgi membranes from mammalian soft tissues by discontinuous density gradient. The degree of Golgi enrichment can be determined by assaying the acitivty of UDP-galactosyl transferase or by immunodetection of Golgi specific marker proteins like B-COP or GM130 using appropriate antibodies (Cat. No. G6160 and G7295, respectively). Separation from other organelles can be measured using the appropriate marker detection kits (Cat. No. CS0780, CYTOCOX1, CY0100 and CAT100).

Application

Golgi Isolation Kit may be used for the isolation of Golgi membranes from mammalian soft tissues by discontinuous density gradient.

Analysis Note

The Golgi Isolation kit was optimized using rat liver and tested on rat kidney, spleen, and heart.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CPISONUC10111667327001
technique(s)

fractionation: suitable

technique(s)

centrifugation: suitable

technique(s)

-

technique(s)

-

usage

sufficient for 50 g (tissue)

usage

-

usage

sufficient for 25 nuclei preparations (~1-10×107 cells/preparation)

usage

-

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution 5 mL
    SDS

Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



V J Allan et al.
The Journal of cell biology, 113(2), 347-359 (1991-04-01)
When higher eukaryotic cells enter mitosis, membrane organization changes dramatically and traffic between membrane compartments is inhibited. Since membrane transport along microtubules is involved in secretion, endocytosis, and the positioning of organelles during interphase, we have explored whether the mitotic
E R Sjoberg et al.
The Journal of biological chemistry, 268(14), 10185-10196 (1993-05-15)
The melanoma-associated disialogangliosides 9(7)-O-acetyl-GD3 and 9(7)-O-acetyl-GD2 have been structurally well characterized. However, the compartmentalization and sequence of action of the biosynthetic activities responsible for synthesizing these molecules remain obscure. Here, we have studied the spatial and temporal interrelationships among the
Julien Villeneuve et al.
The Journal of cell biology, 217(2), 649-665 (2017-12-08)
An appreciation of the functional properties of the cytoplasmic fatty acid binding protein 4 (FABP4) has advanced with the recent demonstration that an extracellular form secreted by adipocytes regulates a wide range of physiological functions. Little, however, is known about



Global Trade Item Number

SKUGTIN
GL0010-1KT04061833654798

Questions

Reviews

No rating value

Active Filters