Skip to Content
Merck

Skip To

G8764

Gc-globulin from human plasma

≥90% (SDS-PAGE), lyophilized powder (containing sodium chloride and sodium phosphate buffer salt)

Synonym(s):

DBP, Group-Specific Component, Vitamin D Binding Protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.61
MDL number:
Form:
lyophilized powder (containing sodium chloride and sodium phosphate buffer salt)
Assay:
≥90% (SDS-PAGE)
Biological source:
human plasma
Mol wt:
~52 kDa
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human plasma

Quality Segment

assay

≥90% (SDS-PAGE)

form

lyophilized powder (containing sodium chloride and sodium phosphate buffer salt)

mol wt

~52 kDa

technique(s)

LC/MS: suitable

solubility

H2O: soluble 2 mg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... GC(2638)

General description

Gc-Globulin, a member of the albumin superfamily, is the major plasma carrier of vitamin D and its metabolites. It is also, along with gelsolin, part of the extracellular actin scavenger system. Gc-globulin also binds monomeric actin (G-actin) with high affinity to scavenge actin released by necrotic cells, preventing its polymerization into filaments. Gc-globulin enhances the chemoattractant activity of C5a for inflammatory cells.
Gc-globulin or the vitamin D binding protein (DBP) is a 51-58 kDa multifunctional serum glycoprotein synthesized in large quantities by primarily in the liver.[1] It is a member of the albumin gene family and has the characteristic multiple disulfide-bonded, triple domain structure.[2]
Gc-globulin from human plasma is encoded by the gene GC, which is mapped to human chromosome 4q13.3.

Application

Gc-globulin is a multifunctional, albumin-like plasma protein that often requires cell surface binding to mediate some of its diverse functions.[3] It has been used to anlayze its glycosylation patterns.
Gc-globulin from human plasma has been used:
  • as a fluorescent probe for G-actin staining
  • as a fluorescent probe to monitor steady-state rate of actin filament disassembly
  • as a calibrator for liquid chromatography with mass spectrometry (LC-MS/MS)

Biochem/physiol Actions

Gc-Globulin, a member of the albumin superfamily, is the major plasma carrier of vitamin D and its metabolites. It is also, along with gelsolin, a part of the extracellular actin scavenger system. Gc-globulin also binds monomeric actin (G-actin) with high affinity to scavenge actin released by necrotic cells, preventing its polymerization into filaments. Gc-globulin enhances the chemoattractant activity of C5a for inflammatory cells.
Single nucleotide polymorphism in the gene GC is associated with deficiency of 25-hydroxyvitamin D [25(OH)D].

Preparation Note

Native Gc-globulin from human plasma

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters