Skip to Content
Merck

Skip To

G154

Gabapentin

≥98% (HPLC), solid, anticonvulsant

Synonym(s):

1-(Aminomethyl)-cyclohexaneacetic acid, Neurontin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C9H17NO2
CAS Number:
Molecular Weight:
171.24
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
262-076-3
MDL number:
Beilstein/REAXYS Number:
2359739
Form:
solid
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Gabapentin, solid

form

solid

Quality Level

color

off-white

solubility

H2O: 10 mg/mL

SMILES string

NCC1(CC(O)=O)CCCCC1

InChI

1S/C9H17NO2/c10-7-9(6-8(11)12)4-2-1-3-5-9/h1-7,10H2,(H,11,12)

InChI key

UGJMXCAKCUNAIE-UHFFFAOYSA-N

Application

Gabapentin has been used:
  • as the α2δ-ligand to examine whether the allyl isothiocyanate (AITC) sensitized thermal aversion assay might be useful as a screening method for the identification of presumptive analgesics[1]
  • dissolved in uptake buffer which is used to measure the to measure ASP (trans-4-[4-(Dimethylamino)styryl]-1-methylpyridinium iodide) substrate by organic cation transporter type 2 (OCTN2) using liquid chromatography - mass spectrometry[2]
  • used as an analgesic adjuvant for intraperitoneal injections

Biochem/physiol Actions

Anticonvulsant with unknown mechanism of action; crosses the blood brain barrier; increases GABA concentrations in the brain and reduces excitatory amino acid neurotransmission, perhaps through its effects on voltage-gated calcium channels; exhibits antinociceptive, anxiolytic, neuroprotective and anti-epileptic effects.
Gabapentin serves as a ligand for the α2δ subunit of L-type calcium channels.[1]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
5.085061287303G-021
form

solid

form

solid

form

-

form

liquid

Quality Level

200

Quality Level

100

Quality Level

-

Quality Level

300

solubility

H2O: 10 mg/mL

solubility

water: 100 mM

solubility

-

solubility

-

color

off-white

color

white

color

-

color

-

Gene Information

human ... ADORA1(134), CACNA1A(773), CACNA1B(774), CACNA1C(775), CACNA1D(776), CACNA1E(777), CACNA1F(778), CACNA1G(8913), CACNA1H(8912), CACNA1I(8911), CACNA1S(779), CACNA2D1(781), CACNA2D2(9254), CACNA2D3(55799), CACNA2D4(93589), CACNB1(782), CACNB2(783), CACNB3(784), CACNB4(785), CACNG1(786), CACNG2(10369), CACNG3(10368), CACNG4(27092), CACNG5(27091), CACNG6(59285), CACNG7(59284), CACNG8(59283)
rat ... Cacna1a(25398)

Gene Information

-

Gene Information

human ... CACNA1A(773), CACNA1B(774), CACNA1C(775), CACNA1D(776), CACNA1E(777), CACNA1F(778), CACNA1G(8913), CACNA1H(8912), CACNA1I(8911), CACNA1S(779), CACNA2D1(781), CACNA2D2(9254), CACNA2D3(55799), CACNA2D4(93589), CACNB1(782), CACNB2(783), CACNB3(784), CACNB4(785), CACNG1(786), CACNG2(10369), CACNG3(10368), CACNG4(27092), CACNG5(27091), CACNG6(59285), CACNG7(59284), CACNG8(59283)

Gene Information

human ... CACNA1A(773), CACNA1B(774), CACNA1C(775), CACNA1D(776), CACNA1E(777), CACNA1F(778), CACNA1G(8913), CACNA1H(8912), CACNA1I(8911), CACNA1S(779), CACNA2D1(781), CACNA2D2(9254), CACNA2D3(55799), CACNA2D4(93589), CACNB1(782), CACNB2(783), CACNB3(784), CACNB4(785), CACNG1(786), CACNG2(10369), CACNG3(10368), CACNG4(27092), CACNG5(27091), CACNG6(59285), CACNG7(59284), CACNG8(59283)


Still not finding the right product?

Explore all of our products under Gabapentin


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



The impact of advanced opioid drugs and analgesic adjuvants on murine macrophage oxygen burst
Kozlowski M, et al.
Folia Medica Cracoviensia (2017)
Thomas R Belliotti et al.
Journal of medicinal chemistry, 48(7), 2294-2307 (2005-04-02)
Pregabalin exhibits robust activity in preclinical assays indicative of potential antiepileptic, anxiolytic, and antihyperalgesic clinical efficacy. It binds with high affinity to the alpha(2)-delta subunit of voltage-gated calcium channels and is a substrate of the system L neutral amino acid
Samuel J Dobson et al.
Antiviral research, 179, 104819-104819 (2020-05-12)
During virus entry, members of the Polyomaviridae transit the endolysosomal network en route to the endoplasmic reticulum (ER), from which degraded capsids escape into the cytoplasm and enter the nucleus. Emerging evidence suggests that viruses require both endosomal acidification and



Global Trade Item Number

SKUGTIN
G154-50MG04061833628263
G154-10MG04061833628256

Questions

  1. Once re-suspended in H20, at what temperature should this be stored at ? I cannot see any specification on this page.

    1 answer
    1. This product is soluble in H₂O at 100 mM. Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

      Helpful?

Reviews

No rating value

Active Filters