Skip to Content
Merck

Skip To

FLAAS

Adenosine 5′-triphosphate (ATP) disodium salt hydrate

≥90%, bacterial (Corynebacterium sp), lyophilized powder

Synonym(s):

ATP disodium hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C10H14N5O13P3Na2 · xH2O
CAS Number:
Molecular Weight:
551.14 (anhydrous basis)
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.26
EC Number:
213-579-1
MDL number:
UNSPSC Code:
12352208
Assay:
≥90%
Biological source:
bacterial (Corynebacterium sp)
Form:
lyophilized powder
Solubility:
water: soluble, clear, colorless (1 vial + 0.1 mL)
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of ~1 mg ATP

biological source

bacterial (Corynebacterium sp)

Quality Level

assay

≥90%

form

lyophilized powder

packaging

vial of ~1 mg ATP

solubility

water: soluble, clear, colorless (1 vial + 0.1 mL)

storage temp.

−20°C

SMILES string

Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O

InChI

1S/C10H16N5O13P3.2Na.H2O/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;;1H2/q;2*+1;/p-2/t4-,6-,7-,10-;;;/m1.../s1

InChI key

NTBQNWBHIXNPRU-MSQVLRTGSA-L

Application

Adenosine 5′-triphosphate (ATP) disodium salt hydrate has been:
  • used as a standard to measure ATP by bioluminescence method[1]
  • used in kinase buffer to assess the interaction between death-associated protein kinases (DAPK2) and 14-3-3-β
  • used as a standard in ATP colorimetric assay

Biochem/physiol Actions

Adenosine 5′-triphosphate was shown to have inhibitory effects on the inflammatory response by abrogating secretion of TNF-α and promoting secretion of Interleukin-10. [2]
P2 purinergic agonist; increases activity of Ca2+-activated K+ channels; substrate for ATP-dependent enzyme systems
Adenosine 5′-triphosphate (ATP) is one of the nucleoside triphosphate present in living system. It controls various biologic processes such as glucose metabolism, muscle contraction and neurotransmission. Apart from this, ATP serves as an important energy source for the cell. Administration of ATP might be useful in the treatment of non-small-cell lung cancer.

Preparation Note

Each vial contains ~1.2 mg of Na2ATP • 3H2O equivalent to ~1.0 mg of ATP (2.0 μmol). Actual amount printed on the label.
Prepared to be substantially vanadium-free.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A1852A3377A2383
biological source

bacterial (Corynebacterium sp)

biological source

microbial (Corynebacterium sp.)

biological source

microbial

biological source

microbial

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

assay

≥90%

assay

≥99% (HPLC)

assay

≥97% (HPLC)

assay

≥99%

form

lyophilized powder

form

powder

form

crystalline

form

powder

packaging

vial of ~1 mg ATP

packaging

vial of 30 mg

packaging

-

packaging

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Els L R Swennen et al.
European journal of immunology, 35(3), 852-858 (2005-02-19)
In vitro studies suggest that extracellular nucleotides and nucleosides may be important regulators of inflammatory and immune responses. Most studies with adenosine 5'-triphosphate (ATP) have been performed in cell lines, which are remote from the human situation. The purpose of
Necrostatin-1 attenuates mitochondrial dysfunction in neurons and astrocytes following neonatal hypoxia--ischemia
Chavez-Valdez R, et al.
Neuroscience, 219, 192-203 (2012)
Francisco J Adrián et al.
Nature chemical biology, 2(2), 95-102 (2006-01-18)
Chronic myelogenous leukemia (CML) is a myeloproliferative disorder characterized at the molecular level by the expression of Bcr-abl, a 210-kDa fusion protein with deregulated tyrosine kinase activity. Encouraged by the clinical validation of Bcr-abl as the target for the treatment



Global Trade Item Number

SKUGTIN
FLAAS-5VL04061833622896
FLAAS-1VL04061833622889
FLAAS-10VL04061838078957

Questions

Reviews

No rating value

Active Filters