Skip to Content
Merck

Skip To

F8666

Lipopolysaccharides from Escherichia coli O55:B5

FITC conjugate

Synonym(s):

LPS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
EC Number:
297-473-0
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Escherichia coli (O55:B5)

Quality Level

conjugate

FITC conjugate

form

lyophilized powder

extent of labeling

2-10 μg FITC per mg LPS

color

yellow to orange

solubility

H2O: 4.90-5.10 mg/mL, faintly hazy to hazy, pale yellow to yellow

shipped in

ambient

storage temp.

2-8°C

General description

This product is a TCA extract of E. coli serotype O55:B5 (strain CDC 1644-70), which was purified by gel filtration and conjugated to FITC (fluorescein isothiocyanate).

Application

FITC-Lipopolysaccharides (FITC-LPS) may be used to study the movement/transport of LPS across tissue barriers such as the colonic epithelium and cellular binding and internalization of lipopolysaccharides with fluorescence-based assay systems.[1][2] FITC-LPS may be used to evaluate, qualify and quantify lipopolysaccharides (LPS) removal reagents and procedures.[3] FITC-LPS conjugate may be used for the detection of LPA binding proteins and motifs.[4]
Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).

Biochem/physiol Actions

Lipopolysaccharides (LPS) are localized in the outer layer of the membrane and are, in noncapsulated strains, exposed on the cell surface. They contribute to the integrity of the outer membrane, and protect the cell against the action of bile salts and lipophilic antibiotics.

Preparation Note

The product is soluble in water (5 mg/ml), yielding a hazy faint yellow solution. Lipopolysaccharides are molecules that form micelles in every solvent. Hazy solutions are observed in water and phosphate buffered saline. Organic solvents do not give clearer solutions. Methanol yields a turbid suspension with floaters, while water yields a homogeneously hazy solution.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F3665L5418L3012
biological source

Escherichia coli (O55:B5)

biological source

Escherichia coli (O111:B4)

biological source

Escherichia coli (O55:B5)

biological source

Escherichia coli (O111:B4)

solubility

H2O: 4.90-5.10 mg/mL, faintly hazy to hazy, pale yellow to yellow

solubility

H2O: soluble 4.90-5.10 mg/mL, faintly hazy to hazy, pale yellow to yellow

solubility

water: 4.90-5.10 mg/mL, faintly hazy to hazy, colorless to faintly yellow

solubility

water: soluble

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

300

form

lyophilized powder

form

lyophilized powder

form

aqueous solution

form

lyophilized powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

color

yellow to orange

color

yellow to orange

color

white to faint yellow

color

white to faint yellow


Still not finding the right product?


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Tatyana Y Postnikova et al.
Pharmaceuticals (Basel, Switzerland), 13(3) (2020-03-22)
Infectious diseases in early postnatal ontogenesis often result in cognitive impairments, particularly learning and memory. The essential foundation of learning and memory is long-term synaptic plasticity, which depends on N-methyl-D-aspartate (NMDA) receptors. In the present study, bacterial infection was modeled
Elena Topchiy et al.
PloS one, 11(5), e0155030-e0155030 (2016-05-14)
Sepsis is the leading cause of death in critically ill patients. While decreased Proprotein Convertase Subtilisin/Kexin type 9 (PCSK9) function improves clinical outcomes in murine and human sepsis, the mechanisms involved have not been fully elucidated. We tested the hypothesis
Yang Yu et al.
Scientific reports, 6, 20845-20845 (2016-02-10)
Phospholipid transfer protein (PLTP) participates in high density lipoprotein (HDL) metabolism. Increased plasma PLTP activity was observed in lipopolysaccharide (LPS) triggered acute inflammatory diseases. This study aimed to determine the exact role of PLTP in LPS induced inflammation. HDL pool



Global Trade Item Number

SKUGTIN
F8666-5MG04061833620939
F8666-10MG04061833620922

Questions

  1. What is the molecular weight?

    1 answer
    1. The molecular weight of this product is not determined. However, LPS is heterogeneous and tends to form aggregates of varying sizes that may range from 1-4 million or greater. When treated with SDS and heat the molecular weight range is approximately 50 to 100 kDa. The FITC used to prepare this product has a molecular weight of 389.38 g/mol. There are approximately 3.6 ug FITC per milligram LPS in this preparation.

      Please see the link below to learn more about Lipopolysaccharides:
      https://www.sigmaaldrich.com/technical-documents/protocol/clinical-testing-and-diagnostics-manufacturing/bacteriology/lipopolysaccharides

      Helpful?

Reviews

No rating value

Active Filters