Skip to Content
Merck

Skip To

F7129

5-Fluorocytosine

nucleoside analog

Synonym(s):

4-amino-5-fluoro-2(1H)-pyrimidinone, Flucytosine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C4H4FN3O
CAS Number:
Molecular Weight:
129.09
UNSPSC Code:
41116107
NACRES:
NA.32
PubChem Substance ID:
EC Number:
217-968-7
Beilstein/REAXYS Number:
127285
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (TLC)

form

powder

mol wt

129.09 g/mol

concentration

≤100% (Flucytosine)

color

white to off-white

mp

298-300 °C (dec.) (lit.)

solubility

formic acid: 50 mg/mL, colorless to faintly yellow

λmax

285-287 nm

fluorescence

(EmM Anhydrous 8.8 - 9.4, pH 1.0)

antibiotic activity spectrum

fungi

mode of action

DNA synthesis | interferes, protein synthesis | interferes

storage temp.

2-8°C

SMILES string

NC1=NC(=O)NC=C1F

InChI

1S/C4H4FN3O/c5-2-1-7-4(9)8-3(2)6/h1H,(H3,6,7,8,9)

InChI key

XRECTZIEBJDKEO-UHFFFAOYSA-N

General description

Chemical structure: nucleoside

Application

Used in studies on TMP biosynthesis.

Biochem/physiol Actions

Nucleoside analog that has antifungal activities. 5-FC is deaminated by cytosine deaminase to product 5-fluorouracil, resulting in RNA miscoding. 5-Fluorocytosine inhibits DNA and RNA synthesis and interferes with ribosomal protein synthesis.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F0175000PHR16591272000
antibiotic activity spectrum

fungi

antibiotic activity spectrum

-

antibiotic activity spectrum

-

antibiotic activity spectrum

-

mode of action

DNA synthesis | interferes, protein synthesis | interferes

mode of action

-

mode of action

-

mode of action

-

Quality Level

200

Quality Level

-

Quality Level

300

Quality Level

-

assay

≥99% (TLC)

assay

-

assay

-

assay

-

form

powder

form

-

form

-

form

-

concentration

≤100% (Flucytosine)

concentration

-

concentration

-

concentration

-


Still not finding the right product?

Explore all of our products under 5-Fluorocytosine


pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
F7129-5G04061833619285
F7129-1G04061835506316

Questions

Reviews

No rating value

Active Filters