Skip to Content
Merck

Skip To

F3685

Fibroblast Growth Factor-Basic, human

≥97% (SDS-PAGE), recombinant, expressed in E. coli, carrier free, suitable for cell culture

Synonym(s):

FGB-b, FGF-2, HBGF-2, HBGH-2, Prostatropin, bFGF

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
Biological source:
human
Recombinant:
expressed in E. coli
Assay:
≥97% (SDS-PAGE)
Form:
lyophilized powder
Mol wt:
16.0 kDa
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fibroblast Growth Factor-Basic, FGF-Basic, from human, recombinant, expressed in E. coli, carrier free

biological source

human

Quality Segment

recombinant

expressed in E. coli

assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

<1 ng/mL Biological Activity EC50

mol wt

16.0 kDa

packaging

pkg of 4X25 μg, pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

color

white to faint yellow cast

solubility

water: soluble 0.025 mg, clear, colorless to faintly yellow

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... FGF2(2247)

General description

Since FGF-Basic is found in a variety of organs, acts on a wide range of cell types, and has multifunctional actions, it has acquired numerous synonyms, including heparin-binding growth factor (class II or beta), eye-derived growth factor I, cartilage-derived growth factor, and astroglial growth factor II5. Purified bovine and human FGF-basic differ by 3 amino acids in sequence3, and are biologically and immunologically cross-reactive.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F5392F5542F7301
assay

≥97% (SDS-PAGE)

assay

~90% (SDS-PAGE and N-terminal analysis)

assay

≥97% (SDS-PAGE)

assay

>95% (SDS-PAGE and HPLC)

recombinant

expressed in E. coli

recombinant

-

recombinant

expressed in E. coli

recombinant

expressed in E. coli

biological source

human

biological source

bovine pituitary glands

biological source

human

biological source

human

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

Gene Information

human ... FGF2(2247)

Gene Information

bovine ... FGF2(281161)

Gene Information

human ... FGF1(2246)

Gene Information

human ... FGF18(8817)


Still not finding the right product?

Explore all of our products under Fibroblast Growth Factor-Basic


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters