Skip to Content
Merck

Skip To

F0132

Anti-Human IgG (γ-chain specific)−FITC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Human IgG (γ-chain specific)−Fluorescein isothiocyanate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
FITC conjugate
Clone:
polyclonal
Application:
IF (d)
Citations:
8
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct immunofluorescence: 1:32

storage temp.

2-8°C

target post-translational modification

unmodified

General description

IgGs are glycoprotein antibodies that regulate immunological defense mechanisms such as phagocytosis and inflammatory responses . IgG also modulates complement-mediated cell lysis . Anti-Human IgG (γ-chain specific) specifically reacts with human IgG when tested against purified human IgM, IgA, IgG, Bence Jones κ, and Lambda myeloma proteins.

Immunogen

human IgG

Application

Anti-Human IgG (γ-chain specific) is suitable for use in indirect immunofluorescence and western immunoblot .

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A3150F5512F3512
biological source

goat

biological source

goat

biological source

goat

biological source

goat

technique(s)

direct immunofluorescence: 1:32

technique(s)

direct ELISA: 1:7,000-1:21,000

technique(s)

immunofluorescence: 1:16-1:32 using Hep2 cells

technique(s)

immunofluorescence: 1:32-1:64 using Hep2 cells

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

conjugate

FITC conjugate

conjugate

alkaline phosphatase conjugate

conjugate

FITC conjugate

conjugate

FITC conjugate

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Thatiana Corrêa de Melo et al.
International journal of molecular sciences, 23(18) (2022-09-24)
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) has been responsible for the severe pandemic of acute respiratory disease, coronavirus disease 2019 (COVID-19), experienced in the 21st century. The clinical manifestations range from mild symptoms to abnormal blood coagulation and severe
C M Shih et al.
Journal of clinical microbiology, 35(2), 450-454 (1997-02-01)
An asymptomatic Babesia infection was confirmed by laboratory diagnoses. The intraerythrocytic protozoan (designed TW1) isolated from a 51-year-old Taiwanese woman appeared to be morphologically consistent with small-form piroplasm, and measurements indicated that it had a body size of 1.5 to
C I Bindon et al.
The Journal of experimental medicine, 168(1), 127-142 (1988-07-01)
Humanized antibodies are likely to have a major role in therapy and it is important to define their interaction with physiological effectors. By comparing a matched series of chimeric human mAbs we found that igG1 was most efficient in complement



Global Trade Item Number

SKUGTIN
F0132-1ML04061833610831
F0132-2ML04061837516283
F0132-.5ML04061833610800

Questions

Reviews

No rating value

Active Filters