Skip to Content
Merck

Skip To

D9938

KIMBLE Dounce tissue grinder set

15 mL complete

Synonym(s):

885300-0015, Dounce grinder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NB.22
UNSPSC Code:
41121800
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

glass

feature

autoclavable

manufacturer/tradename

Kimble® 885300-0015

pestle A clearance

0.0035-0.0065 in.

pestle B clearance

0.0010-0.0030 in.

working volume × L

15 mL × 157 mm

General description

Designed primarily for cellular work where the nucleus remains intact after homogenization.

  • All-glass construction
  • Two pestles are supplied with each complete unit: Large clearance pestle is used for the initial sample reduction; Small clearance pestle is used to form the final homogenate
  • Replacement components are available and completely interchangeable

Features and Benefits

  • Manufactured from borosilicate glass 3.3_x000D_
  • Designed primarily for cellular work where the nucleus remains intact after homogenization._x000D_
  • All-glass construction_x000D_
  • Two pestles are supplied with each complete unit_x000D_
  • Large clearance pestle is used for the initial sample reduction_x000D_
  • Small clearance pestle is used to form the final homogenate_x000D_
  • Replacement components are available and completely interchangeable

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D0189D8938D9063
material

glass

material

glass

material

glass

material

glass

feature

autoclavable

feature

autoclavable

feature

autoclavable

feature

autoclavable

pestle B clearance

0.0010-0.0030 in.

pestle B clearance

0.0005-0.0055 in.

pestle B clearance

0.0005-0.0025 in.

pestle B clearance

0.0008-0.0022 in.

pestle A clearance

0.0035-0.0065 in.

pestle A clearance

0.0020-0.0100 in.

pestle A clearance

0.0030-0.0050 in.

pestle A clearance

0.0028-0.0047 in.

working volume × L

15 mL × 157 mm

working volume × L

100 mL × 232 mm

working volume × L

2 mL × 60 mm

working volume × L

7 mL × 125 mm

manufacturer/tradename

Kimble® 885300-0015

manufacturer/tradename

Kimble® 885300-0100

manufacturer/tradename

Kimble® 885300-0002

manufacturer/tradename

Kimble® 885300-0007


Still not finding the right product?

Explore all of our products under KIMBLE Dounce tissue grinder set




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Emily Petruccelli et al.
Genetics, 215(1), 103-116 (2020-03-07)
Repeated alcohol experiences can produce long-lasting memories for sensory cues associated with intoxication. These memories can problematically trigger relapse in individuals recovering from alcohol use disorder (AUD). The molecular mechanisms by which ethanol changes memories to become long-lasting and inflexible



Global Trade Item Number

SKUGTIN
D9938-1SET04061833575062