Skip to Content
Merck

Skip To

D9050

N-Acetyl-2,3-dehydro-2-deoxyneuraminic acid

≥93% (TLC)

Synonym(s):

2,3-Dehydro-2-deoxy-N-acetylneuraminic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C11H17NO8
CAS Number:
Molecular Weight:
291.25
UNSPSC Code:
12352201
NACRES:
NA.25
PubChem Substance ID:
EC Number:
246-550-7
Beilstein/REAXYS Number:
8722455
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥93% (TLC)

form

powder

optical activity

[α]20/D 40.8 to 51.0 °, c = 0.66% (w/v) in water

color

white

solubility

H2O: soluble 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H]1[C@@H](O)C=C(O[C@H]1[C@H](O)[C@H](O)CO)C(O)=O

InChI

1S/C11H17NO8/c1-4(14)12-8-5(15)2-7(11(18)19)20-10(8)9(17)6(16)3-13/h2,5-6,8-10,13,15-17H,3H2,1H3,(H,12,14)(H,18,19)/t5-,6+,8+,9+,10+/m0/s1

InChI key

JINJZWSZQKHCIP-UFGQHTETSA-N

Application

N-Acetyl-2,3-dehydro-2-deoxyneuraminic acid (NADNA), a specific endogenous neuraminidase (NEU) inhibitor, may be used to study the roles of endogenous neuraminidase in the development and function of neural processes and pathways as well as other processes that depend upon sialylation-desialylation cycles. [1][2]

Biochem/physiol Actions

Inhibitor of bacterial, viral and animal neuraminidase (sialidase).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
110138D1133T2144
assay

≥93% (TLC)

assay

≥98% (HPLC)

assay

≥98% (titration)

assay

≥93% (HPLC)

biological source

synthetic (organic)

biological source

-

biological source

synthetic (organic)

biological source

-

solubility

H2O: soluble 50 mg/mL, clear, colorless

solubility

water: ≥10 mg/mL

solubility

1 M HCl: 50 mg/mL, clear, colorless

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

Quality Level

300

Quality Level

100

Quality Level

300

Quality Level

200

form

powder

form

solid

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

room temp

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
D9050-5MG04061833590188
D9050-10MG04061833590171
D9050-25MG04061832389240

Questions

Reviews

No rating value

Active Filters