Skip to Content
Merck

Skip To

D8016

Sodium 2,3-dimercaptopropanesulfonate monohydrate

95%

Synonym(s):

2,3-Dimercaptopropanesulfonic acid sodium salt monohydrate, DMPS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
HSCH2CH(SH)CH2SO3Na · H2O
CAS Number:
Molecular Weight:
228.29
UNSPSC Code:
12161900
NACRES:
NA.25
PubChem Substance ID:
EC Number:
223-796-3
Beilstein/REAXYS Number:
3734863
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

zwitterionic

Quality Level

assay

95%

form

powder

mol wt

228.29 g/mol

mp

215 °C (dec.)

storage temp.

2-8°C

SMILES string

O.[Na+].[O-]S(=O)(=O)CC(S)CS

InChI

1S/C3H8O3S3.Na.H2O/c4-9(5,6)2-3(8)1-7;;/h3,7-8H,1-2H2,(H,4,5,6);;1H2/q;+1;/p-1

InChI key

XMUHNMQFDVIWGU-UHFFFAOYSA-M

General description

Sodium 2,3-dimercaptopropanesulfonate monohydrate is a chelating agent for heavy metals. [1]

Application

Sodium 2,3-dimercaptopropanesulfonate monohydrate has been used in a study to assess its use as an antidote against non-metallic pesticides as nereistoxin insecticides. It has also been noted in a review of existing chelating agents for the treatment of heavy metals and metalloid intoxication. [2]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
30968431648172553
form

powder

form

powder

form

crystals and lumps

form

powder

assay

95%

assay

≥99.0% (NT)

assay

≥99.98% trace metals basis

assay

97%

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

-

mp

215 °C (dec.)

mp

-

mp

-

mp

>300 °C (lit.)

mol wt

228.29 g/mol

mol wt

average mol wt 1200-5000

mol wt

-

mol wt

-

description

zwitterionic

description

anionic

description

-

description

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Sodium dimercaptopropane sulfonate as antidote against non-metallic pesticides
Chen, Z., et al.
Acta Pharmacologica Sinica, 25, 534-534 (2001)
Santiago Iglesia-Turiño et al.
Plant physiology, 142(2), 742-749 (2006-08-22)
The mercury (Hg) accumulation mechanism was studied in rape (Brassica napus) plants grown under a Hg concentration gradient (0 microm-1,000 microm). Hg mainly accumulated in roots. Therefore, the presence of phytochelatins (PCs) was studied in the roots of the plants.
Swaran J S Flora et al.
International journal of environmental research and public health, 7(7), 2745-2788 (2010-08-19)
Chelation therapy is the preferred medical treatment for reducing the toxic effects of metals. Chelating agents are capable of binding to toxic metal ions to form complex structures which are easily excreted from the body removing them from intracellular or



Global Trade Item Number

SKUGTIN
D8016-1G04061826690635
D8016-250MG04061833270578

Questions

Reviews

No rating value

Active Filters