Skip to Content
Merck

Skip To

D7170

2′-Deoxyguanosine 5′-triphosphate trisodium salt solution

10 mM in H2O

Synonym(s):

dGTP-Na3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H13N5Na3O13P3
CAS Number:
Molecular Weight:
573.13
NACRES:
NA.52
PubChem Substance ID:
UNSPSC Code:
41106305
EC Number:
300-026-5
MDL number:
Assay:
≥99%
Form:
liquid
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99%

Quality Level

form

liquid

concentration

10 mM in H2O

color

colorless

foreign activity

DNase, RNase, NICKase, none detected

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].NC1=Nc2c(ncn2[C@H]3C[C@H](O)[C@@H](COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)O3)C(=O)N1

InChI

1S/C10H16N5O13P3.3Na/c11-10-13-8-7(9(17)14-10)12-3-15(8)6-1-4(16)5(26-6)2-25-30(21,22)28-31(23,24)27-29(18,19)20;;;/h3-6,16H,1-2H2,(H,21,22)(H,23,24)(H2,18,19,20)(H3,11,13,14,17);;;/q;3*+1/p-3/t4-,5+,6+;;;/m0.../s1

InChI key

IWGGLKOTEOCWQP-BIHLCPNHSA-K

General description

2′-Deoxyguanosine 5′-triphosphate (dGTP) is made of a guanine nucleobase attached to deoxyribose and a 5′-hydroxyl on the sugar linked to a chain of three phosphate residues. It is used by the cells for the synthesis of DNA with the help of DNA polymerase.

Application

2′-Deoxyguanosine 5′-triphosphate trisodium salt is utilized for methods such as DNA sequencing, PCR and other DNA polymerase based DNA synthesis and repair techniques.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
DGTP-ROD503811051466001
Quality Level

200

Quality Level

-

Quality Level

200

Quality Level

-

form

liquid

form

solution

form

liquid

form

solution

assay

≥99%

assay

99% (dNTP)

assay

≥99%

assay

98%

color

colorless

color

-

color

colorless

color

-

concentration

10 mM in H2O

concentration

<0.9% (dNDP), 100 mM

concentration

100 mM (pH 7)

concentration

100 mM

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Michael G Mohsen et al.
Nucleic acids research, 47(18), 9495-9501 (2019-09-11)
We document the preparation and properties of dimerized pentaphosphate-bridged deoxynucleotides (dicaptides) that contain reactive components of two different nucleotides simultaneously. Importantly, dicaptides are found to be considerably more stable to hydrolysis than standard dNTPs. Steady-state kinetics studies show that the



Global Trade Item Number

SKUGTIN
D7170-.5ML04061838214478

Questions

Reviews

No rating value

Active Filters