Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352202
NACRES:
NA.25
Pricing and availability is not currently available.
Product Name
6% SigMatrix Serum diluent,
biological source
synthetic
Quality Segment
description
Buffered Recombinant Human Serum Albumin
shipped in
wet ice
storage temp.
2-8°C
General description
This reagent is a 6% recombinant HSA (Human Serum Albumin) in PBS solution, pH 7.4, that can be used as a blank or matrix/diluent for proteinaceous analytes in LC-MS/MS calibrators, controls, or samples.The 6% protein content design is meant to mimic endogenous protein concentration in human serum samples.Beneficially, as the HSA source is recombinant, the matrix should not contain any measurable contaminants typically found in even stripped serum.
Physical form
The product contains 6% recombinant human serum albumin in a PBS solution containing 0.01% ProClin™ 950 as a preservative.
Preparation Note
For more information on other Sigmatrix products visit the Sigmatrix Selection Guide.
This solution is provided ready to use as a 6% (w/v) Recombinant Human Serum Albumin in Phosphate Buffered Saline, pH 7.4. This formulation has been shown to be appropriate for use as a sample diluent or matrix for preparing calibrants/controls for LC-MS/MS assays.
Legal Information
ProClin is a trademark of LANXESS Corp.
1 of 1
This Item | |||
|---|---|---|---|
| description Buffered Recombinant Human Serum Albumin | description - | description - | description synthetic negative urine control |
| biological source synthetic | biological source - | biological source human | biological source synthetic |
| Quality Level 200 | Quality Level - | Quality Level 300 | Quality Level 200 |
| shipped in wet ice | shipped in - | shipped in wet ice | shipped in wet ice |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C |
Still not finding the right product?
Explore all of our products under 6% SigMatrix Serum diluent
Storage Class
12 - Non Combustible Liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


