Skip to Content
Merck

Skip To

D4818

Dispase® I

protease

Synonym(s):

Protease from Bacillus polymyxa

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
EC Number:
255-914-4
NACRES:
NA.54
MDL number:
Specific activity:
≥10 unit/mg solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

lyophilized solid

specific activity

≥10 unit/mg solid

technique(s)

cell culture | mammalian: suitable, single cell analysis: suitable

storage temp.

2-8°C

Application

Dispase I has been used in a study to assess the effect of amniotic membrane on wound size in the early stages of the healing process. Dispase I has also been used in a study to investigate a dityrosine-based substrate for a protease assay.
Dispase I has been used in lung digestion and processing for flow staining, as well as for CD4 cell isolation in mice.[1] The enzyme has also been used to digest excised wounds and a small amount of surrounding skin for the detection of GFP+ (green fluorescence protein) cells. This study to investigated the effect of differentiation and angiogenesis of bone marrow-derived mesenchymal stem cells on wound healing.[2] It has also been used to remove the epidermis during the isolation of dermal fibroblasts from mice.[3]

Suitable for use in preparation of single cell suspension for sequencing.

Biochem/physiol Actions

Dispase I is a rapid, effective, gentle and neutral protease that can separate intact epidermis from the dermis. It can also separate intact epithelial sheets in culture from the substratum. The enzyme preserves the viability of the epithelial cells while cleaving the basement membrane zone region. It can also be used to prevent clumping in suspension cultures. This protease cleaves fibronectin and type IV collagen, but not laminin, type V collagen, serum albumin, or transferrin.[4] It hydrolyzes N-terminal peptide bonds of non-polar amino acid residues. It preferentially attacks denatured and intercellular proteins with exposed hydrophobic amino acid residues. Ca2+, Mg2+, Mn2+, Fe2+, Fe3+ and Al3+ activate the enzyme. EDTA, EGTA, Hg2+ and other heavy metals inhibit the enzyme activity.[5] The enzyme contains 1g-atom of zinc per g-mol of purified enzyme. If this zinc component is removed by chelating agents such as EDTA or EGTA, an inactive apoenzyme is obtained. The enzyme is not inhibited by serum.

Physical form

lyophilized powder containing calcium acetate

Other Notes

One unit will hydrolyze casein to produce color equivalent to 1.0 μmole (181 μg) of tyrosine per min at pH 7.5 at 37 °C (color by Folin-Ciocalteu reagent), unless otherwise indicated.

Legal Information

Dispase is a registered trademark of Godo Shusei Co., Ltd.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D4693T7902C9891
technique(s)

cell culture | mammalian: suitable, single cell analysis: suitable

technique(s)

cell culture | mammalian: suitable, single cell analysis: suitable

technique(s)

cell culture | mammalian: suitable, single cell analysis: suitable

technique(s)

cell culture | mammalian: suitable, single cell analysis: suitable

specific activity

≥10 unit/mg solid

specific activity

≥0.5 units/mg solid

specific activity

30-350 units/mg protein (E1%/280)

specific activity

≥125 CDU/mg solid, 0.5-5.0 FALGPA units/mg solid

form

lyophilized solid

form

lyophilized solid

form

powder

form

essentially salt-free, lyophilized powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Alizée Vercauteren Drubbel et al.
Cell stem cell, 28(8), 1411-1427 (2021-04-22)
Columnar metaplasia of the esophagus is the main risk factor for esophageal adenocarcinoma. There is a lack of evidence to demonstrate that esophageal progenitors can be the source of columnar metaplasia. In this study, using transgenic mouse models, lineage tracing
Yaojiong Wu et al.
Stem cells (Dayton, Ohio), 25(10), 2648-2659 (2007-07-07)
Although chronic wounds are common, treatment for these disabling conditions remains limited and largely ineffective. In this study, we examined the benefit of bone marrow-derived mesenchymal stem cells (BM-MSCs) in wound healing. Using an excisional wound splinting model, we showed
Marco Tatullo et al.
Materials (Basel, Switzerland), 12(4) (2019-02-20)
Human periapical cyst mesenchymal stem cells (hPCy-MSCs) are a newly discovered cell population innovatively collected from inflammatory periapical cysts. The use of this biological waste guarantees a source of stem cells without any impact on the surrounding healthy tissues, presenting



Global Trade Item Number

SKUGTIN
D4818-2MG04061833585719

Questions

1–6 of 6 Questions  
  1. How to disolve dispase? Can I use mediums as DMEM, PBS ...?

    1 answer
    1. The enzyme may be reconstituted at 1 mg/ml in 10 mM sodium acetate buffer, pH 7.5 with 5 mM calcium acetate. Historical data also note that this material may be reconstituted in PBS or other physiologic buffers containing Ca2+, culture medium, or water; however, no concentration is provided.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. At what concentration should Product D4818, Dispase, be used?

    1 answer
    1. To dissociate cells from ECM (extracellular matrix) use at a concentration of 0.6 - 2.4 units/ml.

      Helpful?

  4. What is the solubility of Product D4818, Dispase I?

    1 answer
    1. The enzyme may be reconstituted at 1 mg/ml in 10 mM sodium acetate buffer, pH 7.5 with 5 mM calcium acetate.

      Helpful?

  5. What is the difference between Product D4693, Dispase II  and Product D4818, Dispase I?

    1 answer
    1. Product D4693 is a lyophilized powder containing calcium acetate and milk sugar as a stabilizer. The activity specification is >0.5 unit/mg solid. Product D4818 does not contain any milk sugar and has an activity specification of >10 units/mg solid.

      Helpful?

  6. What is the activity of the Product D4818, Dispase, which I received?

    1 answer
    1. Each lot will have a specific activity that can be found on our website on the certificate of analysis.

      Helpful?

Reviews

No rating value

Active Filters