Skip to Content
Merck

Skip To

D1069

Desloratadine

≥98% (HPLC), histamine H1 receptor antagonist, powder

Synonym(s):

8-Chloro-6,11-dihydro-11-(4-piperidinylidene)-5H- benzo[5,6]cyclohepta[1,2,b]pyridine, Clarinex®, NSC 675447

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H19ClN2
CAS Number:
Molecular Weight:
310.82
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Desloratadine, powder, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Clc1ccc2c(CCc3cccnc3\C2=C4\CCNCC4)c1

InChI

1S/C19H19ClN2/c20-16-5-6-17-15(12-16)4-3-14-2-1-9-22-19(14)18(17)13-7-10-21-11-8-13/h1-2,5-6,9,12,21H,3-4,7-8,10-11H2

InChI key

JAUOIFJMECXRGI-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Application

Desloratadine has been used to test its effect on embryoid body development in in vitro gastrulation model of P19C5 stem cells. It has also been used as an antihistamine mimic in rats and to treat lung cancer (A549) and glioblastoma (U87) cells in various cellular studies.

Biochem/physiol Actions

A non-sedating H1-receptor antagonist, free from antimuscarinic/anticholinergic effects. It has novel antiallergic and anti-inflammatory effects.
Desloratadine is a selective and nonsedating histamine H1 receptor antagonist, an active metabolite of loratadine (Claritin), used to relieve hay fever and allergy symptoms with less drowsiness than other antihistamines; does not significantly inhibits cardiac K+ channels at clinically achievable blood levels. Free from antimuscarinic/anticholinergic effects.

Legal Information

Clarinex is a registered trademark of Schering Corp.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR16801173042Y0001584
assay

≥98% (HPLC)

assay

-

assay

-

assay

-

Quality Level

100

Quality Level

300

Quality Level

-

Quality Level

-

form

powder

form

-

form

-

form

-

storage temp.

2-8°C

storage temp.

2-30°C

storage temp.

-

storage temp.

2-8°C

Gene Information

human ... HRH1(3269)

Gene Information

human ... HRH1(3269)

Gene Information

human ... HRH1(3269)

Gene Information

human ... HRH1(3269)

solubility

DMSO: >10 mg/mL

solubility

-

solubility

-

solubility

-


Still not finding the right product?

Explore all of our products under Desloratadine


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

390.2 °F - Pensky-Martens closed cup

flash_point_c

199 °C - Pensky-Martens closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
D1069-50MG04061833558522
D1069-10MG04061833558515

Questions

Reviews

No rating value

Active Filters