Skip to Content
Merck

Skip To

D0390

NG,NG′-Dimethyl-L-arginine di(p-hydroxyazobenzene-p′-sulfonate) salt

≥99% (TLC)

Synonym(s):

SDMA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H18N4O2 · 2C12H10N2O4S
CAS Number:
Molecular Weight:
758.82
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥99% (TLC)

form

powder

solubility

methanol: 10 mg/mL

storage temp.

room temp

SMILES string

CN\C(NC)=N\CCC[C@H](N)C(O)=O.Oc1ccc(cc1)\N=N\c2ccc(cc2)S(O)(=O)=O.Oc3ccc(cc3)\N=N\c4ccc(cc4)S(O)(=O)=O

InChI

1S/2C12H10N2O4S.C8H18N4O2/c2*15-11-5-1-9(2-6-11)13-14-10-3-7-12(8-4-10)19(16,17)18;1-10-8(11-2)12-5-3-4-6(9)7(13)14/h2*1-8,15H,(H,16,17,18);6H,3-5,9H2,1-2H3,(H,13,14)(H2,10,11,12)/b2*14-13+;/t;;6-/m..0/s1

InChI key

HISUXCNNXYEHER-YJSASASGSA-N

Application

Plasma sym-dimethylarginine has been considered a marker of hepatic and renal dysfunction. A sensitive LC-MS method for identifying arginine, and both asym- and sym-dimethylarginine in plasma and urine samples has been developed.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
724041D426837966
assay

≥99% (TLC)

assay

95% (CP)

assay

≥98% (TLC)

assay

≥97% (TLC)

biological source

synthetic (organic)

biological source

-

biological source

-

biological source

synthetic

Quality Level

200

Quality Level

-

Quality Level

200

Quality Level

100

form

powder

form

powder

form

powder

form

powder or crystals

storage temp.

room temp

storage temp.

−20°C

storage temp.

room temp

storage temp.

room temp

solubility

methanol: 10 mg/mL

solubility

-

solubility

water: 50 mg/mL, clear, colorless

solubility

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters