Skip to Content
Merck

Skip To

C9394

Chlorhexidine digluconate solution

20% in H2O

Synonym(s):

Bis(p-chlorophenyl)diguanidohexane digluconate, 1,6-Bis(N5-[p-chlorophenyl]-N1-biguanido)hexane; 1,1′-Hexamethylenebis(5-[p-chlorophenyl]biguanide)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
NACRES:
NA.85
PubChem Substance ID:
UNSPSC Code:
51102829
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Level

concentration

20% in H2O

solubility

water: soluble 50 % (w/v)

density

1.06 g/mL at 25 °C (lit.)

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.Clc1ccc(NC(=N)NC(=N)NCCCCCCNC(=N)NC(=N)Nc2ccc(Cl)cc2)cc1

InChI

1S/C22H30Cl2N10.2C6H12O7/c23-15-5-9-17(10-6-15)31-21(27)33-19(25)29-13-3-1-2-4-14-30-20(26)34-22(28)32-18-11-7-16(24)8-12-18;2*7-1-2(8)3(9)4(10)5(11)6(12)13/h5-12H,1-4,13-14H2,(H5,25,27,29,31,33)(H5,26,28,30,32,34);2*2-5,7-11H,1H2,(H,12,13)/t;2*2-,3-,4+,5-/m.11/s1

InChI key

YZIYKJHYYHPJIB-UUPCJSQJSA-N

Application

Chlorhexidine has been used to study how essential oils improve skin antisepsis when combined with chlorhexidine digluconate[1] and is used for skin permeation studies[2] . Chronic rinsing with chlorhexidine has been shown to decrease the saltiness of NaCl and the bitterness of quinine[3].
Used to study mechanism of membrane dysruption by bis(biguanide) molecules.

Biochem/physiol Actions

Cationic broad-spectrum antimicrobial agent belonging to the bis(biguanide) family. Its mechanism of action involves destabilization of the outer bacterial membrane. It is used primarily as a topical antiseptic/disinfectant in wound healing, at catheterization sites, in various dental applications and in surgical scrubs.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light sensitive.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C6143PHR1294282227
mode of action

cell membrane | interferes

mode of action

cell membrane | interferes

mode of action

-

mode of action

-

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

antibiotic activity spectrum

-

antibiotic activity spectrum

-

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

100

form

liquid

form

powder

form

-

form

powder

storage temp.

2-8°C

storage temp.

room temp

storage temp.

2-8°C

storage temp.

-

concentration

20% in H2O

concentration

-

concentration

-

concentration

-


pictograms

CorrosionEnvironment

signalword

Danger

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Product Information Sheet


T J Karpanen et al.
Antimicrobial agents and chemotherapy, 52(10), 3633-3636 (2008-08-05)
This study evaluated a model of skin permeation to determine the depth of delivery of chlorhexidine into full-thickness excised human skin following topical application of 2% (wt/vol) aqueous chlorhexidine digluconate. Skin permeation studies were performed on full-thickness human skin using
Chandra C Shenoy et al.
Endocrinology, 158(10), 3605-3619 (2017-09-25)
Progressive scarring is ubiquitous postoperatively and in an array of chronic systemic diseases. Recent studies indicate that such scarring has a high female propensity; females are also almost exclusively affected by endometriosis, a common sex steroid-dependent fibrotic disease. Endometriosis-related fibrosis
P A Breslin et al.
Chemical senses, 26(2), 105-116 (2001-03-10)
Chronic rinsing with chlorhexidine, an oral-antiseptic, has been shown to decrease the saltiness of NaCl and the bitterness of quinine. The effect of acute chlorhexidine on taste has not been investigated. The purpose of the present study was to examine



Global Trade Item Number

SKUGTIN
C9394-25ML04061838255075
C9394-100ML04061838255068
C9394-500ML04061838255082

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Hi, Is this pharma grade ? Kidn regards Archana

    1 answer
    1. No, this product is not pharmaceutical grade. It is intended for research use only. Currently, a pharmaceutical grade of this material is not available.

      Helpful?

Reviews

No rating value

Active Filters