Skip to Content
Merck

Skip To

C9377

Chenodeoxycholic acid

Synonym(s):

3α,7α-Dihydroxy-5β-cholanic acid, 5β-Cholanic acid-3α,7α-diol, Chenodiol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57
UNSPSC Code:
12352106
NACRES:
NA.25
PubChem Substance ID:
EC Number:
207-481-8
Beilstein/REAXYS Number:
3219887
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

anionic

form

powder

mol wt

392.57 g/mol

technique(s)

protein quantification: suitable

CMC

(3)

mp

165-167 °C (lit.)

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20-,22+,23+,24-/m1/s1

InChI key

RUDATBOHQWOJDD-BSWAIDMHSA-N

Gene Information

General description

Chenodeoxycholic acid is a bile acid synthesized in the liver from cholesterol.

Application

Chenodeoxycholic acid has been used in a study to assess its effects as a long-term replacement therapy for cerebrotendinous xanthomatosis (CTX). [1] It has also been used in a study to investigate its effects on the small-intestinal absorption of bile acids in patients with ileostomies. [2]

Biochem/physiol Actions

Bile Acid

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
700198PC1050000BP926
technique(s)

protein quantification: suitable

technique(s)

-

technique(s)

-

technique(s)

-

form

powder

form

powder

form

-

form

solid

storage temp.

room temp

storage temp.

−20°C

storage temp.

-

storage temp.

2-8°C

CMC

(3)

CMC

-

CMC

-

CMC

-

mp

165-167 °C (lit.)

mp

-

mp

165-167 °C (lit.)

mp

165-167 °C (lit.)

mol wt

392.57 g/mol

mol wt

-

mol wt

-

mol wt

-


Still not finding the right product?

Explore all of our products under Chenodeoxycholic acid


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



M O Osman et al.
Gut, 43(2), 232-239 (1999-04-06)
Interleukin 8 (IL-8) has recently been proposed to have an important role in mediating the development of the systemic sequelae associated with severe acute pancreatitis. To define the role of IL-8 in acute pancreatitis by neutralising its effects with a
Gallstone dissolution in man using chenodeoxycholic acid
Bell, G., et al.
Lancet, 300, 1213-1213 null
A Stiehl et al.
Gastroenterology, 98(2), 424-428 (1990-02-01)
The effects of ursodeoxycholic acid and chenodeoxycholic acid on the small-intestinal absorption of endogenous bile acids were studied in patients with ileostomies who served as a model to investigate small-intestinal absorption in humans. In the control period, the eight patients



Global Trade Item Number

SKUGTIN
C9377-25G04061833530566
C9377-100MG04061833530559
C9377-5G04061833530573

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters