Skip to Content
Merck

Skip To

C8707

Cystamine dihydrochloride

BioXtra

Synonym(s):

2,2′-Diaminodiethyl disulfide dihydrochloride, 2,2′-Dithiobis(ethylamine) dihydrochloride, Decarboxycystine dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
NH2CH2CH2SSCH2CH2NH2 · 2HCl
CAS Number:
Molecular Weight:
225.20
UNSPSC Code:
12352200
NACRES:
NA.25
PubChem Substance ID:
EC Number:
200-260-7
Beilstein/REAXYS Number:
3616850
MDL number:
Assay:
≥98% (TLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioXtra

Quality Level

assay

≥98% (TLC)

form

powder

impurities

≤0.0005% Phosphorus (P), ≤0.1% Insoluble matter

ign. residue

≤0.1%

mp

217-220 °C (dec.) (lit.)

solubility

H2O: 0.1 M, clear, colorless to light yellow

anion traces

sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%, Ca: ≤0.02%, Cu: ≤0.0005%, Fe: ≤0.001%, K: ≤0.005%, Mg: ≤0.0005%, NH4+: ≤0.05%, Na: ≤0.005%, Pb: ≤0.001%, Zn: ≤0.0005%

SMILES string

Cl[H].Cl[H].NCCSSCCN

InChI

1S/C4H12N2S2.2ClH/c5-1-3-7-8-4-2-6;;/h1-6H2;2*1H

InChI key

YUFRRMZSSPQMOS-UHFFFAOYSA-N

Application

Cystamine dihydrochloride is a useful reagent to derivatize various polymer monoliths for hydrophilic interaction liquid chromatography; as a crosslinking agent in the development of polymer hydrogels; and as a functional group in nanoparticles developed for siRNA and DNA delivery.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
108318PHR927030050
assay

≥98% (TLC)

assay

-

assay

-

assay

≥98.0% (AT)

Quality Level

200

Quality Level

500

Quality Level

300

Quality Level

200

solubility

H2O: 0.1 M, clear, colorless to light yellow

solubility

-

solubility

-

solubility

water: soluble 1 g/10 mL, clear, colorless

form

powder

form

solid

form

-

form

powder

mp

217-220 °C (dec.) (lit.)

mp

217-220 °C (dec.) (lit.)

mp

217-220 °C (dec.) (lit.)

mp

217-220 °C (dec.) (lit.), ~220 °C (dec.)

anion traces

sulfate (SO42-): ≤0.05%

anion traces

-

anion traces

-

anion traces

-


Still not finding the right product?

Explore all of our products under Cystamine dihydrochloride


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Liang Gao et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(11), 3185-3192 (2012-02-22)
Autofluorescent microcapsules were assembled by covalent cross-linking of polysaccharide alginate dialdehyde (ADA) derivative and cystamine dihydrochloride (CM) through a layer-by-layer (LBL) technique. The formulated Schiff base and disulfide bonds render capsules with pH- and redox-responsive properties for pinpointed intracellular delivery
Jie Gao et al.
Langmuir : the ACS journal of surfaces and colloids, 28(9), 4464-4471 (2012-01-27)
Gold nanoparticles (GNPs) are attractive alternative optical probes and good biocompatible materials due to their special physical and chemical properties. However, GNPs have a tendency to aggregate particularly in the presence of high salts and certain biological molecules such as
Thorsten Overath et al.
Molecular & cellular proteomics : MCP, 11(8), 467-477 (2012-05-05)
The post-translational modification of proteins with O-GlcNAc is involved in various cellular processes including signal transduction, transcription, translation, and nuclear transport. This transient protein modification enables cells or tissues to adapt to nutrient conditions or stress. O-Glycosylation of the 26



Global Trade Item Number

SKUGTIN
C8707-100G04061833619186
C8707-25G04061833610336

Questions

Reviews

No rating value

Active Filters