Skip to Content
Merck

Skip To

C8649

Cholesterol Oxidase from Streptomyces sp.

lyophilized powder, ≥20 units/mg protein

Synonym(s):

Cholesterol: oxygen oxidoreductase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-842-1
MDL number:
EC Number:
Specific activity:
≥20 units/mg protein
Biological source:
Streptomyces sp.
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Streptomyces sp.

Quality Segment

form

lyophilized powder

specific activity

≥20 units/mg protein

mol wt

~34 kDa

composition

protein, 55-65% biuret

solubility

50 mM potassium phosphate buffer, pH 7.0: soluble (Cold)

shipped in

dry ice

storage temp.

−20°C

General description

Isoelectric point : 5.1 ± 0.1 and 5.4 ± 0.1
Michaelis constant : 4.3 x 10‾5M(Cholesterol)
Inhibitors : Ionic detergents, Hg++, Ag+
Optimum pH : 6.5 − 7.0
Optimum temperature : 45 – 50°C
pH Stability : pH 5.0 – 10.0 (25°C, 20hr)
Thermal stability : Below 45°C (pH 7.0, 15min)
Cholesterol Oxidase produced from Streptomyces sp corresponds to a molecular weight of 62 kDa. It has a pH and temperature optimum of 7 and 37 °C, respectively.[1] Cholesterol Oxidase comprises a FAD-binding and a steroid-binding domain.

Application

Cholesterol oxidase from Streptomyces has been used in a study to assess the relationship between the micellar structure of model bile and the activity of esterase. Cholesterol oxidase from Streptomyces has also been used in a study to investigate the effects of sphingomyelin degradation on cell cholesterol oxidizability and steady-state distribution between the cell surface and the cell interior.
Cholesterol oxidase is used to determine serum cholesterol.[2][3] The enzyme also finds application in the microanalysis of steroids in food samples and in distinguishing 3-ketosteroids from 3β-hydroxysteroids.[4] Transgenic plants expressing cholesterol oxidase are being investigated in the fight against the cotton boll weevil.[5] CHOD has also been used as a molecular probe to elucidate cellular membrane structures.[6][7]

Biochem/physiol Actions

Cholesterol oxidase (CHOD) is a monomeric flavoprotein containing FAD that catalyzes the first step in cholesterol catabolism. This bifunctional enzyme oxidizes cholesterol to cholest-5-en-3-one in an FAD-requiring step, which is then isomerized to cholest-4-en-3-one with the release of H2O2.[8]
Cholesterol Oxidase from microorganisms use cholesterol for carbon and energy. It is useful in diagnostic assays involving cholesterol, especially in lipid disorders. Cholesterol oxidase has an insecticidal property and aids protection against boll weevil larvae. The Streptomyces sp cholesterol oxidase has long shelf-life with good performance.

Physical form

lyophilized powder containing bovine serum albumin and amino acids as stabilizers

Preparation Note

CHOD is soluble in cold 50 mM potassium phosphate buffer, pH 7.0. Prepare solutions immediately before use.

Other Notes

One unit will convert 1.0 μmole of cholesterol to 4-cholesten-3-one per min at pH 7.5 at 25 °C. Note: 4-cholesten-3-one may undergo isomerization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
228250C5421C1235
biological source

Streptomyces sp.

biological source

-

biological source

-

biological source

-

specific activity

≥20 units/mg protein

specific activity

≥3 units/mg solid, ≥40 U/mg

specific activity

≥30 units/mg protein (biuret)

specific activity

≥10 units/mg protein

form

lyophilized powder

form

lyophilized solid

form

aqueous solution

form

lyophilized powder

mol wt

~34 kDa

mol wt

-

mol wt

62 kDa

mol wt

55 kDa

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−70°C

storage temp.

−20°C

solubility

50 mM potassium phosphate buffer, pH 7.0: soluble (Cold)

solubility

water: 20 mg/mL

solubility

50 mM potassium phosphate buffer, pH 7.0: soluble (Cold)

solubility

50 mM potassium phosphate buffer, pH 7.0: soluble


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
C8649-250UN04061833522653
C8649-100UN04061833522646

Questions

Reviews

No rating value

Active Filters