Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
CL124-1H6, monoclonal
form
buffered aqueous solution
mol wt
antigen ~27 kDa
species reactivity
human
concentration
~2 mg/mL
technique(s)
indirect ELISA: suitable, western blot: 10-20 μg/mL using total cell extracts of HCT-116 cells
isotype
IgG2b
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CTSK(1513)
General description
Immunogen
Application
Biochem/physiol Actions
Physical form
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| species reactivity human | species reactivity mouse, mink, human, rat | species reactivity human | species reactivity human |
| biological source mouse | biological source mouse | biological source rabbit | biological source mouse |
| clone CL124-1H6, monoclonal | clone 33/2, monoclonal | clone polyclonal | clone CTD-19, monoclonal |
| Gene Information human ... CTSK(1513) | Gene Information human ... CTSL1(1514) | Gene Information human ... CTSK(1513) | Gene Information human ... CTSD(1509) |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form purified immunoglobulin | antibody form ascites fluid | antibody form affinity isolated antibody | antibody form ascites fluid |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C8243-200UL | 04061838030504 |
| C8243-100UL | 04061838030498 |



