Skip to Content
Merck

Skip To

C7912

Cefotaxime sodium salt

potency: 916-964 μg per mg

Synonym(s):

Cefotaxime, Cefotaxim sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C16H16N5NaO7S2
CAS Number:
Molecular Weight:
477.45
UNSPSC Code:
51284136
NACRES:
NA.85
PubChem Substance ID:
EC Number:
264-915-9
Beilstein/REAXYS Number:
5711411
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

potency

916-964 μg per mg

solubility

H2O: 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].[H][C@]12SCC(COC(C)=O)=C(N1C(=O)[C@H]2NC(=O)C(=N/OC)\c3csc(N)n3)C([O-])=O

InChI

1S/C16H17N5O7S2.Na/c1-6(22)28-3-7-4-29-14-10(13(24)21(14)11(7)15(25)26)19-12(23)9(20-27-2)8-5-30-16(17)18-8;/h5,10,14H,3-4H2,1-2H3,(H2,17,18)(H,19,23)(H,25,26);/q;+1/p-1/b20-9-;/t10-,14-;/m1./s1

InChI key

AZZMGZXNTDTSME-JUZDKLSSSA-M

General description

Chemical structure: β-lactam

Application

Cefotaxim has been used to find new residues involved in cefotaxime hydrolysis in CTX-M β-lactamases,[1] to study antibiotic-susceptible and -resistant Streptococcus pneumoniae infections,[2] and to study pneumococcal pneumonia and the pharmokinetics of various treatments.[3] It may be used to study the effects of binding and inhibition of penicillin binding protein 2 (PBP2) on bacterial mucopeptide synthesis.

Biochem/physiol Actions

Cefotaxim inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs) which inhibits the final transpeptidation step of peptidoglycan synthesis in bacterial cell walls. As a result, bacteria lyse due to cell wall autolytic enzymes.
Mode of Action: Inhibits bacterial cell wall synthesis.

Other Notes

Broad spectrum third generation cephalosporin antibiotic.
Keep container tightly closed in a dry and well-ventilated place.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C70391097909PHR2099
antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

antibiotic activity spectrum

-

antibiotic activity spectrum

-

Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

300

form

powder

form

powder or crystals

form

-

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

potency

916-964 μg per mg

potency

-

potency

-

potency

-

solubility

H2O: 50 mg/mL

solubility

H2O: 10% ( )

solubility

-

solubility

-


Still not finding the right product?

Explore all of our products under Cefotaxime sodium salt


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Product Information Sheet






Global Trade Item Number

SKUGTIN
C7912-5G04061833521359
C7912-1G04061833521342

Questions

Reviews

No rating value

Active Filters