Skip to Content
Merck

Skip To

C7512

Butyrylcholinesterase from equine serum

lyophilized powder, ≥10 units/mg protein

Synonym(s):

Acylcholine acyl-hydrolase, Choline esterase, butyryl, Pseudocholinesterase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-579-2
MDL number:
EC Number:
Specific activity:
≥10 units/mg protein
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized powder

Quality Level

specific activity

≥10 units/mg protein

mol wt

tetramer 440 kDa

composition

Protein, ≥60%

solubility

cold water: 60 U/mL

storage temp.

−20°C

General description

Butyrylcholinesterase (BChE) is a serine hydrolase that is structurally similar to acetylcholinesterase (AChE), but differs in substrate specificities and inhibitor sensitivities. The enzyme is a tetrameric glycoprotein with four equal subunits (110 kDa each). It is also termed as pseudocholinesterase. It is produced in the liver. It is found in blood, the synapse of neuromuscular junctions and glia cells and axons of white matter.

Application

Butyrylcholinesterase from equine serum has been used in a microcalorimetric study of the inhibition of butyrylcholinesterase by paraoxon. This enzyme has also been used in a study to investigate the synthesis and inhibition of cholinergic enzymes. Selective inhibition of BChE activity can be used in the detection of organophosphates. Its use in the treatment of organophosphate toxicity has shown clinical potential, as there is a correlation between the blood level of BChE in humans and the degree of protection against toxic nerve agents.[1] There has also been an interest in the roles of cholinesterases with regard to Alzheimer′s disease. Investigations into selective inhibitors may provide a clearer picture of the physiological role of BChE in both healthy and diseased individuals.[2] This product has been used for the screening of cholineesterase inhibitors in selected fruits and vegetables, for restoring cognitive function and improving memory. It has also been used to develop a butyrylcholinesterase and choline oxidase immobilized bio-sniffer for the detection of nicotine. Nicotine inhibits BChE activity. A decrease in the byproducts of BChE activity reflects the volume of nicotine.[3]

Biochem/physiol Actions

Butyrylcholinesterase (BChE) can, unlike AChE, efficiently hydrolyze larger esters of choline such as butyrylcholine and benzoylcholine. The enzyme is activated by Ca2+ and Mg2+ and the activity is constant over the pH range 6.0-8.0. It is inhibited by betaine, nicotine, organophosphates, and carbamates.
Mutations in butyrylcholinesterase (BChE) gene results in the failure of respiratory system, prolonged paralysis and apnea.

Physical form

Lyophilized powder containing buffer salts

Preparation Note

This product is prepared from equine serum using ammonium sulfate fractionation and supplied as a lyophilized powder.

Analysis Note

Protein determined by biuret

Other Notes

One unit will hydrolyze 1.0 μmole of butyrylcholine to choline and butyrate per min at pH 8.0 at 37 °C. The activity obtained using butyrylcholine as substrate is ~2.5 times that obtained using acetylcholine.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C1057C4290B4186
specific activity

≥10 units/mg protein

specific activity

≥900 units/mg protein

specific activity

≥500 units/mg protein

specific activity

≥50 units/mg protein

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

powder

solubility

cold water: 60 U/mL

solubility

H2O: soluble 10 mg/mL

solubility

-

solubility

-

mol wt

tetramer 440 kDa

mol wt

tetramer 440 kDa

mol wt

-

mol wt

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

200


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Toxicokinetics of Chemical Warfare Agents: Nerve Agents and Vesicants
Handbook of Toxicology of Chemical Warfare Agents, 755-790 (2009)
Kohji Mitsubayashi et al.
Analytica chimica acta, 573-574, 69-74 (2007-08-29)
A novel bioelectronic sniffer for nicotine in the gas phase was developed with enzyme inhibition principle to butyrylcholinesterase activity. The bioelectronic devices for nicotine in the gas and liquid phases were constructed using a Clark-type dissolved oxygen electrode and a
Screening for cholinesterase inhibitors in selected fruits and vegetables.
Szwajgier D and Borowiec K
Biotechnology, 15(2) (2012)



Global Trade Item Number

SKUGTIN
C7512-12KU04061832755366
C7512-6KU04061832755373
C7512-1.2KU04061833512579

Questions

  1. I want to buy product C7512 but i will encapsulate the BChE and the app. weight size is more important for me. But I can only see the IU value of these products. Can you inform me the app. weight of BChE in each three of products? Thanks.

    1 answer
    1. The actual amount of powder in each vial is lot-specific. This product has a minimum protein concentration of 60% and a minimum activity specification of 10 units per milligram of protein. For example, a 1200-unit package for a lot reporting 60% protein and 14 units per milligram of protein, will contain approximately 142 milligrams per vial.
      Please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/344/841/c7512dat.pdf

      Please see the link below to review the enzymatic assay protocol:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/323/602/c7512enz.pdf

      Please see the link below to review a sample or lot specific Certificate:
      https://www.sigmaaldrich.com/product/sigma/c7512*product-documentation

      Helpful?

Reviews

No rating value

Active Filters