Skip to Content
Merck

Skip To

C7268

Chymostatin

powder

Synonym(s):

N-(Nα-Carbonyl-Cpd-X-Phe-al)-Phe (Cpd = capreomycidine) (capreomycidine = [S,S]-α-(2-Iminohexahydro-4-pyrimidyl)glycine)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Chymostatin, microbial

biological source

microbial

Quality Segment

form

powder

solubility

DMSO: 10 mM (Stock solutions stable for months at −20 °C.)

storage temp.

−20°C

SMILES string

OC(C(NC(NC(C1NC(NCC1)=N)C([F,Cl,Br,I]C)=O)=O)CC2=CC=CC=C2)=O

InChI key

MRXDGVXSWIXTQL-HYHFHBMOSA-N

Application

Chymostatin has been used in a study that determined that molecular calculations are useful for evaluating the interactions between ligands, including inhibitors and homologous enzymes, in docking models. Chymostatin has also been used in a study to investigate the norovirus protease as an attractive target for antiviral drug development.

Biochem/physiol Actions

Chymostatin is a strong inhibitor of many proteases, including chymotrypsin, papain, chymotrypsin-like serine proteinases, chymases, and lysosomal cysteine proteinases such as cathepsins A,B,C, B, H, and L. It weakly inhibits human leucocyte elastase. It is effective at a final concentration of 100 to 200 μg/ml (10 to 100 μM). Chymostatin is often included in protease inhibitor cocktails used with plant extracts.
Inhibits the lysosomal proteinase cathepsin B, and the soluble Ca2+-activated proteinase.[1] Many young plant tissues express a chomostatin-sensitive serine protease.[2]

Preparation Note

Solubility testing in glacial acetic acid at 10 mg/ml yields a clear solution, which is usually colorless, but can be yellow in appearance. It is reportedly also soluble in DMSO; only slightly soluble in water and short-chain alcohols; insoluble in ethyl acetate, butyl acetate, ether, hexane, and petroleum ether. Stock solutions (10 mM) can be prepared in DMSO and are stable for months at -20 °C. Stock solutions can also be made in 0.1 M HCl. Dilute solutions (10-100 μM) are only stable for several hours, due to oxidation of the terminal aldehyde.

Other Notes

A mixture of A (major), B and C components. A: X=Leu; B: X=Val; C: X=Ile
chymostatin A MW = 607.7
chymostatin B MW = 593.7
chymostatin C MW = 607.7

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
23079011004638001T4376
biological source

microbial

biological source

-

biological source

-

biological source

synthetic

Quality Level

300

Quality Level

100

Quality Level

-

Quality Level

200

form

powder

form

powder, solid

form

powder

form

powder

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

solubility

DMSO: 10 mM (Stock solutions stable for months at −20 °C.)

solubility

5% glacial acetic acid: 10 mg/mL, DMSO: 20 mg/mL

solubility

acetic acid: soluble 20 mg/mL

solubility

DMSO: >10 mg/mL (stable for several months at 4°C.), ethanol: 20 mg/mL


Still not finding the right product?

Explore all of our products under Chymostatin


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters