Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
natural (organic)
Quality Level
product line
BioReagent
form
powder
technique(s)
cell culture | insect: suitable
SMILES string
O1[C@H](C(C(C(C1CO)O)O)O)O[C@@H]2C(OC(C(C2O)O)O)CO
InChI
1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1
InChI key
GUBGYTABKSRVRQ-WFVLMXAXSA-N
General description
Application
- As a component of nutrient-rich diet, to study how nutrition affects individual fitness and insecticide efficacy among German cockroaches.
- As a carbon source for Schizophyllum commune culture.
- To study the effect of cellulose on the content of minerals in the femur of rats.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) cell culture | insect: suitable | technique(s) LPLC: suitable | technique(s) LPLC: suitable | technique(s) LPLC: suitable |
| biological source natural (organic) | biological source - | biological source spruce | biological source - |
| Quality Level 200 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| form powder | form powder | form powder | form powder |
| product line BioReagent | product line - | product line - | product line - |
semi-solid matricies, gels, hydrogels, gel electrophoresis, hydrophilic gels | - | - | - |
Still not finding the right product?
Explore all of our products under α-Cellulose
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C6429-5KG | 04061833511237 |
| C6429-500G | 04061833511220 |
