Skip to Content
Merck

Skip To

C6154

Z-Gln-Gly

γ-glutamyl donor substrate

Synonym(s):

N2-[(phenylmethoxy)carbonyl]-L-glutaminyl-glycine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H19N3O6
CAS Number:
Molecular Weight:
337.33
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352209
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

powder

storage temp.

−20°C

SMILES string

NC(=O)CCC(NC(=O)OCc1ccccc1)C(=O)NCC(O)=O

InChI

1S/C15H19N3O6/c16-12(19)7-6-11(14(22)17-8-13(20)21)18-15(23)24-9-10-4-2-1-3-5-10/h1-5,11H,6-9H2,(H2,16,19)(H,17,22)(H,18,23)(H,20,21)

InChI key

SOUXAAOTONMPRY-UHFFFAOYSA-N

Application

γ-Glutamyl donor substrate used in spectrophotometric determination of transglutaminase (TGase) activity.[1][2] Z-Gln-Gly was used to enzymatically synthesize N-linked neoglycoproteins.[3]

Biochem/physiol Actions

N-Benzyloxycarbonyl-L-Glutaminylglycine (Z-Gln-Gly, Z-QG) is used as a substrate to differentiate and characterize transglutaminase(s) (TGase) that catalyzes the post-translational covalent cross-linking of Gln- and Lys-containing peptides. Z-QG supports glutamyl-level cross-linking applications thruough surface modification.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P3501A0878P4510
form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Natalie M Rachel et al.
Protein science : a publication of the Protein Society, 26(11), 2268-2279 (2017-09-01)
Microbial transglutaminase (MTG) is a practical tool to enzymatically form isopeptide bonds between peptide or protein substrates. This natural approach to crosslinking the side-chains of reactive glutamine and lysine residues is solidly rooted in food and textile processing. More recently
Evan A Wells et al.
Archives of biochemistry and biophysics, 643, 57-61 (2018-02-27)
The Ca2+-dependent deamidation and transamidation activities of transglutaminase 2 (TG2) are important to numerous physiological and pathological processes. Herein, we have examined the steady-state kinetics and 15(V/K) kinetic isotope effects (KIEs) for the TG2-catalyzed deamidation and transamidation of N-Benzyloxycarbonyl-l-Glutaminylglycine (Z-Gln-Gly)
Momoko Kitaoka et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(19), 5387-5392 (2011-04-07)
A new synthetic strategy for DNA-enzyme conjugates with a novel architecture was explored using a natural cross-linking catalyst, microbial transglutaminase (MTG). A glutamine-donor substrate peptide of MTG was introduced at the 5-position on the pyrimidine of deoxyuridine triphosphate to prepare



Global Trade Item Number

SKUGTIN
C6154-1G04061833510872

Questions

  1. Qual è la massima solubilità in H2O del composto Z-Gln-Gly cod.C6154?

    1 answer
    1. This product is tested for solubility at 50 mg/mL in 1 M NaOH. The solubility in water has not been determined.

      Helpful?

Reviews

No rating value

Active Filters