Skip to Content
Merck

Skip To

C5929

Calcium carbonate

BioReagent, suitable for insect cell culture, ≥99.0%

Synonym(s):

Limestone, calcite, calofort u, chalk, marble

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
UNSPSC Code:
12352207
NACRES:
NA.71
PubChem Substance ID:
EC Number:
207-439-9
Beilstein/REAXYS Number:
8008338
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Level

assay

≥99.0%

form

powder

technique(s)

cell culture | insect: suitable

solubility

H2O: soluble (10 g + 20 mL of HCl, dilute to 150 mL with H2O)

density

2.93 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

General description

Calcium carbonate (CaCO3) is a ternary ionic compound formed by carbon, oxygen, and calcium. It is one of the most common minerals found in rocks worldwide. It aids in studying the process of biomineralization, where it participates in the formation of various mineral structures including marine sediments, shells of marine organisms, snails, coal balls, pearls, and eggshells. CaCO3 exists in different mineral forms such as calcite, aragonite, and vaterite.

Application

Calcium carbonate buffer has been used as a component of alginate solution for tissue culture, and for preparing conductive alginate-coated electrodes.[1] It has also been used in the preparation of alginate hydrogel for drug delivery.

Biochem/physiol Actions

Calcium carbonate aids in bone formation and mineralization. It is often used as an antacid due to its potent alkalizing properties. Calcium carbonate has good biocompatibility, porous structure, large surface area, and high loading capacity. These properties of calcium carbonate make it favorable to use as a drug delivery system. In addition, the lower solubility of calcium carbonate (CaCO3) makes it effective in reducing the gelation rate and prolonging the working time for alginate gels. Therefore, it is widely used in the preparation of alginate solution.[2]

Features and Benefits

  • BioReagent grade.
  • High level of assay ( ≥99.0%), indicating its purity.
  • Designed for insect cell culture applications.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
398101239216795445
assay

≥99.0%

assay

99.95-100.05% dry basis

assay

≥99.0%

assay

≥99%

technique(s)

cell culture | insect: suitable

technique(s)

-

technique(s)

-

technique(s)

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

product line

BioReagent

product line

-

product line

-

product line

Redi-Dri

form

powder

form

powder

form

powder

form

powder

density

2.93 g/mL at 25 °C (lit.)

density

2.93 g/mL at 25 °C (lit.)

density

2.93 g/mL at 25 °C (lit.)

density

2.93 g/mL at 25 °C (lit.)


Still not finding the right product?

Explore all of our products under Calcium carbonate


Storage Class

13 - Non Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Calcium Carbonate
Kristina Fritz
StatPearls [Internet] (2023)
The potential of biomimesis in bone tissue engineering: lessons from the design and synthesis of invertebrate skeletons
Green D, et al.
Bone, 30(6) , 810-815 (2002)
Injectable polysaccharide hydrogel embedded with hydroxyapatite and calcium carbonate for drug delivery and bone tissue engineering
Ren B, et al.
International Journal of Biological Macromolecules, 118(Pt A), 1257-1266 (2018)



Global Trade Item Number

SKUGTIN
C5929-100G04061833502730
C5929-500G04061833502815

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters