Skip to Content
Merck
All Photos(3)

Documents

C4282

Sigma-Aldrich

Coenzyme A hydrate

≥85% (UV, HPLC)

Synonym(s):

CoA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H36N7O16P3S · xH2O
CAS Number:
Molecular Weight:
767.53 (anhydrous basis)
Beilstein:
77809
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

biological source

yeast

Assay

≥85% (UV, HPLC)

form

powder

functional group

phospholipid

shipped in

ambient

storage temp.

−20°C

SMILES string

O.CC(C)(COP(O)(=O)OP(O)(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP(O)(O)=O)n2cnc3c(N)ncnc23)[C@@H](O)C(=O)NCCC(=O)NCCS

InChI

1S/C21H36N7O16P3S.H2O/c1-21(2,16(31)19(32)24-4-3-12(29)23-5-6-48)8-41-47(38,39)44-46(36,37)40-7-11-15(43-45(33,34)35)14(30)20(42-11)28-10-27-13-17(22)25-9-26-18(13)28;/h9-11,14-16,20,30-31,48H,3-8H2,1-2H3,(H,23,29)(H,24,32)(H,36,37)(H,38,39)(H2,22,25,26)(H2,33,34,35);1H2/t11-,14-,15-,16+,20-;/m1./s1

InChI key

TVSAELAFGDOPKI-BLPRJPCASA-N

Application

Coenzyme A hydrate has been used in the thiolase enzyme assay of recombinant acetoacetyl-CoA thiolase (rACAT) in Clonorchis sinensis. It may be used as a reference standard in Raman spectra measurements.

Biochem/physiol Actions

Coenzyme A (CoA) is an essential metabolic cofactor synthesized from cysteine, pantothenate, and ATP. CoA plays important roles in many metabolic pathways, including the tricarboxylic acid cycle, and the synthesis and oxidation of fatty acids. One of the main functions of CoA is the carrying and transfer of acyl groups. Acylated deriviates, for example acetyl-CoA, are critical intermediates in many metabolic reactions. CoA levels can be altered during starvation, and in conditions such as cancer, diabetes, and alcoholism.

Caution

The free acid is less stable than the sodium or lithium salt; 5% decomposition may occur within 6 months when stored at −80 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francis McCoy et al.
Molecular cell, 52(3), 325-339 (2013-10-08)
Active metabolism regulates oocyte cell death via calcium/calmodulin-dependent protein kinase II (CaMKII)-mediated phosphorylation of caspase-2, but the link between metabolic activity and CaMKII is poorly understood. Here we identify coenzyme A (CoA) as the key metabolic signal that inhibits Xenopus
Takuya Ishibashi et al.
Extremophiles : life under extreme conditions, 16(6), 819-828 (2012-09-04)
We have previously reported that the majority of the archaea utilize a novel pathway for coenzyme A biosynthesis (CoA). Bacteria/eukaryotes commonly use pantothenate synthetase and pantothenate kinase to convert pantoate to 4'-phosphopantothenate. However, in the hyperthermophilic archaeon Thermococcus kodakarensis, two
Gregory R Wagner et al.
The Journal of biological chemistry, 288(40), 29036-29045 (2013-08-16)
Alterations in mitochondrial protein acetylation are implicated in the pathophysiology of diabetes, the metabolic syndrome, mitochondrial disorders, and cancer. However, a viable mechanism responsible for the widespread acetylation in mitochondria remains unknown. Here, we demonstrate that the physiologic pH and
Haruyuki Atomi et al.
Biochemical Society transactions, 41(1), 427-431 (2013-01-30)
CoA is a ubiquitous molecule in all three domains of life and is involved in various metabolic pathways. The enzymes and reactions involved in CoA biosynthesis in eukaryotes and bacteria have been identified. By contrast, the proteins/genes involved in CoA
John C Newman et al.
The Journal of biological chemistry, 287(51), 42436-42443 (2012-10-23)
The sirtuins are a family of NAD(+)-dependent protein deacetylases that regulate cell survival, metabolism, and longevity. Three sirtuins, SIRT3-5, localize to mitochondria. Expression of SIRT3 is selectively activated during fasting and calorie restriction. SIRT3 regulates the acetylation level and enzymatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service